Skip to Content
Merck
All Photos(1)

Key Documents

SML2846

Sigma-Aldrich

Tebipenem pivoxil

≥98% (HPLC)

Synonym(s):

(4R,5S,6S)-pivaloyloxymethyl 3-(1-(4,5-dihydrothiazol-2-yl)azetidin-3-ylthio)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate, L 084, L-084, L084, ME1211, SPR 994, SPR-994, SPR994, TBM-PI, Tebi-pivoxil, [4R-[4a,5ß,6ß(R*)]]-3-[[1-(4,5-dihydro-2-thiazolyl)-3-azetidinyl]thio]-6-(1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid (2,2-dimethyl-1-oxopropoxy)methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H31N3O6S2
CAS Number:
Molecular Weight:
497.63
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

S1CCN=C1N2CC(C2)SC3=C(N4[C@H]([C@H]3C)[C@H](C4=O)[C@H](O)C)C(=O)OCOC(=O)C(C)(C)C

InChI

1S/C22H31N3O6S2/c1-11-15-14(12(2)26)18(27)25(15)16(19(28)30-10-31-20(29)22(3,4)5)17(11)33-13-8-24(9-13)21-23-6-7-32-21/h11-15,26H,6-10H2,1-5H3/t11-,12-,14-,15-/m1/s1

InChI key

SNUDIPVBUUXCDG-QHSBEEBCSA-N

Biochem/physiol Actions

Orally bioavailable pivalyl ester prodrug form of the carbapenem class broad-spectrum β-lactam antibiotic Tebipenem (LJC 11,036; SPR859).
Tebipenem pivoxil (L-084I; ME1211; SPR994; Tebi-pivoxil; TBM-PI) corresponds to the orally bioavailable pivalyl ester prodrug form of the carbapenem class broad-spectrum β-lactam antibiotic Tebipenem (LJC 11,036; SPR859; TBM). TBM is potent against both gram-positive and gram-negative bacteria, including penicillin-resistant S. pneumoniae (PRSP) and many β-lactamase-producing strains, while being less effective against methicillin-resistant S. aureus (MRSA), S. marcescens, and P. aeruginosa. TBM is 2- to 64-fold more potent than imipenem, cefdinir, and faropenem against clinical isolates from respiratory and urinary-tract infections.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akash Jain et al.
Expert review of anti-infective therapy, 16(7), 513-522 (2018-07-18)
Infections caused by antibiotic-resistant pathogens, particularly Gram-negative bacteria, have become increasingly challenging to successfully treat. The beta-lactam antibiotic subclass, the carbapenems, have proven valuable for the treatment of such Gram-negative bacterial infections due to their spectrum and β-lactamase stability properties.
Muneki Hotomi et al.
Vaccine, 25(13), 2478-2484 (2006-10-24)
An animal model of otitis media using chinchillas was developed to evaluate the efficacy of tebipenem pivoxil (TBM-PI) against experimental otitis media. Chinchillas inoculated via the transbullar approach with Streptococcus pneumoniae serogroup 6 were included in the efficacy study with
Takeshi Isoda et al.
The Journal of antibiotics, 59(4), 241-247 (2006-07-13)
We discovered an orally active carbapenem, L-084, through pharmacokinetic studies on various prodrug esters of (1R,5S,6S)-6-[(R)-1-hydroxyethyl]-l-methyl-2-[1-(1,3-thiazolin-2-yl)azetidin-3-yl]thio-1-carbapen-2-em-3-carboxylic acid (LJC11,036). L-084 showed a strong antimicrobial activity against Gram-positive and Gram-negative bacteria and exhibited the highest intestinal absorption among synthesized prodrugs of LJC11,036.
Visanu Thamlikitkul et al.
Journal of the Medical Association of Thailand = Chotmaihet thangphaet, 97(12), 1259-1268 (2015-03-15)
To determine in vitro and in vivo activity of tebipenem against ESBL-producing E. coli. Minimum inhibitory concentration (MIC) of tebipenem against 100 clinical isolates of ESBL-producing E. coli was performed by broth micro-dilution technique. Blood and urine samples from 10
Kazuhiko Kato et al.
Molecular pharmaceutics, 7(5), 1747-1756 (2010-08-26)
Tebipenem pivoxil (TBPM-PI) is an oral carbapenem antibiotic for treating otolaryngologic and respiratory infections in pediatric patients. This agent is a prodrug to improve intestinal absorption of TBPM, an active form, and an absorption rate of TBPM-PI is higher than

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service