Skip to Content
Merck
All Photos(3)

Key Documents

S8170

Sigma-Aldrich

Sodium nitrate

BioXtra, ≥99.0%

Synonym(s):

Chile salpeter

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NaNO3
CAS Number:
Molecular Weight:
84.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
Assay:
≥99.0%
solubility:
H2O: 1 M at 20 °C, clear, colorless

product line

BioXtra

Quality Level

Assay

≥99.0%

impurities

<0.0005% Phosphorus (P)
<0.1% Insoluble matter

mp

306 °C (dec.) (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): <0.05%
sulfate (SO42-): <0.05%

cation traces

Al: <0.0005%
Ca: <0.0005%
Cu: <0.0005%
Fe: <0.0005%
K: <0.01%
Mg: <0.0005%
NH4+: <0.1%
Pb: <0.001%
Zn: <0.0005%

SMILES string

[Na+].[O-][N+]([O-])=O

InChI

1S/NO3.Na/c2-1(3)4;/q-1;+1

InChI key

VWDWKYIASSYTQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium nitrate is synthesized by treating nitrite with nitric acid. The proportion of nitrite production is higher when synthesized by absorbing waste gases from nitric acid. Supplementation of sodium nitrate may serve to improve acute heart failure (AHF) by modulating the nitric oxide precursor levels.

Application

Sodium nitrate has been used:
  • in combination with beef extract for virus elution from water samples
  • as an external calibrant in nitrate assay to quantify nitrate in cell culture medium
  • as a component of liquid BG11-medium for culturing Synechococcus elongatus PCC 7942
  • to test its protective effects in hepatic ischemia-reperfusion injury

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of Chlorophyll as a Proxy for Biofilm Formation in the Cyanobacterium Synechococcus elongatus
Sendersky E, et al.
Bio-protocol, 7, e2406-e2406 (2017)
Methylglyoxal evokes acute Ca2+ transients in distinct cell types and increases agonist-evoked Ca2+ entry in endothelial cells via CRAC channels
Sachdeva R, et al.
Cell Calcium, 78(3), 66-75 (2019)
Re-evaluation of sodium nitrate (E 251) and potassium nitrate (E 252) as food additives
Mortensen A, et al.
EFSA Journal, 15(6), e04787-e04787 (2017)
Inorganic nitrate as a treatment for acute heart failure: a protocol for a single center, randomized, double-blind, placebo-controlled pilot and feasibility study
Falls R, et al.
Journal of Translational Medicine, 15(1), 172-172 (2017)
Cytoprotective effects of nitrite during in vivo ischemia-reperfusion of the heart and liver
Duranski MR, et al.
The Journal of Clinical Investigation, 115(5), 1232-1240 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service