Skip to Content
Merck
All Photos(3)

Key Documents

S2252

Sigma-Aldrich

Sodium nitrite

ReagentPlus®, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaNO2
CAS Number:
Molecular Weight:
69.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
form:
powder or crystals

grade

reagent

Quality Level

product line

ReagentPlus®

Assay

≥99.0%

form

powder or crystals

autoignition temp.

914 °F

pH

9 (20 °C, 100 g/L)

mp

271 °C (lit.)

SMILES string

[Na+].[O-]N=O

InChI

1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1

InChI key

LPXPTNMVRIOKMN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium nitrite is an air-sensitive and hygroscopic solid with both reducing and oxidizing properties. It is commonly used as a reagent and catalyst in synthetic organic chemistry. It is decomposed by acids (even weak acids) with the evolution of a brown mixture of nitrogen oxides (NOx).

Application

Sodium nitrite can be used as a reagent in:      
  • In the conversion of amines into diazo compounds, which are used as key precursors to many dyes, such as diazo dyes.      
  • Nef reaction and Abidi transformations.      
  • The C–C bond cleavage reaction in the presence of TFA under aerobic conditions.      
  • The oxidative carbonitration of alkenes in the presence of K2S2O8.

It can also be used as a nitrating agent in organic synthesis.
It is used in direct C-H nitration, ipso-nitration, and nitration via transition metal-catalyzed cross-coupling reaction. NaNO2 is also used as a co-catalyst for catalytic oxidation of alcohols under aerobic, solvent-free conditions.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raquel Morais de Paiva Daibert et al.
PloS one, 15(11), e0241861-e0241861 (2020-11-07)
Macrophages are classified upon activation as classical activated M1 and M2 anti-inflammatory regulatory populations. This macrophage polarization is well characterized in humans and mice, but M1/M2 profile in cattle has been far less explored. Bos primigenius taurus (taurine) and Bos
Anne C Rios et al.
Cancer cell, 35(4), 618-632 (2019-04-02)
Breast tumors are inherently heterogeneous, but the evolving cellular organization through neoplastic progression is poorly understood. Here we report a rapid, large-scale single-cell resolution 3D imaging protocol based on a one-step clearing agent that allows visualization of normal tissue architecture
Lucas C Pinheiro et al.
Free radical biology & medicine, 53(4), 701-709 (2012-06-23)
The new pathway nitrate-nitrite-nitric oxide (NO) has emerged as a physiological alternative to the classical enzymatic pathway for NO formation from l-arginine. Nitrate is converted to nitrite by commensal bacteria in the oral cavity and the nitrite formed is then
Edward H Oldfield et al.
Journal of neurosurgery, 119(3), 634-641 (2013-05-28)
Intravenous sodium nitrite has been shown to prevent and reverse cerebral vasospasm in a primate model of subarachnoid hemorrhage (SAH). The present Phase IIA dose-escalation study of sodium nitrite was conducted to determine the compound's safety in humans with aneurysmal
Rustem F Dautov et al.
Nitric oxide : biology and chemistry, 40, 36-44 (2014-05-27)
Previous studies in non-human blood vessels and in platelets have demonstrated that under hypoxic conditions release of NO from nitrite (NO2(-)) is potentiated by deoxyhaemoglobin. In the current study, we characterized hypoxic potentiation of NO2(-) effects in human vasculature and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service