Skip to Content
Merck
All Photos(3)

Key Documents

P8291

Sigma-Aldrich

Potassium nitrate

BioReagent, suitable for cell culture, suitable for plant cell culture

Synonym(s):

KNO3, saltpeter

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KNO3
CAS Number:
Molecular Weight:
101.10
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
grade:
reagent
form:
crystals
solubility:
H2O: 20/150 g/mL, clear, colorless
Pricing and availability is not currently available.

grade

reagent

product line

BioReagent

form

crystals

technique(s)

cell culture | mammalian: suitable
cell culture | plant: suitable

mp

334 °C (lit.)

solubility

H2O: 20/150 g/mL, clear, colorless

SMILES string

[K+].[O-][N+]([O-])=O

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1

InChI key

FGIUAXJPYTZDNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Potassium nitrate has been used:
  • in CPW13M solution to extract the protoplasts from plant cells[1]
  • as a component in the basal medium[2]
  • as a component in macronutrient[3]

Biochem/physiol Actions

Potassium nitrate serves as a source of nitrate for cell growth. It also induces the synthesis of lipid in alga Neochloris oleoabundans.[4]

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biomolecule delivery into canola protoplasts by centrifuging cells with microbubbles
He C, et al.
Febs Letters, , number=(587), 285?290-285?290 (2013)
Bioassay to Study the Attachment of Xanthomonas albilineans on Sugarcane Leaves.
Mensi I, et al.
Bio-protocol, 7(587), 285?290-285?290 (2016)
Extraction and Activity of O-acetylserine(thiol)lyase (OASTL) from Microalga Chlorella sorokiniana
Salbitani G and Carfagna S
Plant & Cell Physiology, , number=(587), 285?290-285?290 (2016)
Effects of nitrogen sources on cell growth and lipid accumulation of green alga Neochloris oleoabundans
Li Y, et al.
Applied Microbiology and Biotechnology, , number=(81), 629?636-629?636 (2008)
Wolfgang R Engelsberger et al.
The Plant journal : for cell and molecular biology, 69(6), 978-995 (2011-11-09)
Nitrogen is an essential macronutrient for plant growth and development. Inorganic nitrogen and its assimilation products control various metabolic, physiological and developmental processes. Although the transcriptional responses induced by nitrogen have been extensively studied in the past, our work here

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service