Skip to Content
Merck
All Photos(1)

Documents

PHR1582

Supelco

Dehydroacetic acid

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Dehydroacetic acid, 2-Acetyl-5-hydroxy-3-oxo-4-hexenoic acid δ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O4
CAS Number:
Molecular Weight:
168.15
Beilstein:
6129
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1166309

API family

dehydroacetic acid

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

270 °C (lit.)

mp

111-113 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CC(=O)C1C(=O)OC(C)=CC1=O

InChI

1S/C8H8O4/c1-4-3-6(10)7(5(2)9)8(11)12-4/h3,7H,1-2H3

InChI key

PGRHXDWITVMQBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dehydroacetic acid is a pyrone derivative, that is typically synthesized by the base-catalyzed dimerization of diketene. It is used as a preservative in pharmaceutical, food and cosmetic products.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Dehydroacetic acid may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical, food and cosmetic formulations by chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA4498 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F

Flash Point(C)

157 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of sorbic acid, dehydroacetic acid and benzoic acid by gas chromatography-mass spectrometry
Kakemoto M
Journal of Chromatography A, 594(1-2), 253-257 (1992)
Comparing micellar electrokinetic chromatography and microemulsion electrokinetic chromatography for the analysis of preservatives in pharmaceutical and cosmetic products
Huang HY, et al.
Journal of Chromatography A, 993(1-2), 153-164 (2003)
Dehydroacetic acid, triacetic acid lactone, and related pyrones
Advances in Heterocyclic Chemistry, 53, 1-84 (1992)
Simultaneous determination of nine kinds of preservatives in foods by HPLC
Kishi H and Yamada T
Journal of Food Hygiene and Safety, 48(3), 58-63 (2007)
E Mikami et al.
Journal of pharmaceutical and biomedical analysis, 28(2), 261-267 (2002-04-04)
A high-performance liquid chromatographic (HPLC) method for simultaneous determination of dehydroacetic acid (DHA), benzoic acid (BA), sorbic acid (SOA) and salicylic acid (SA) was developed for application to cosmetic products. Isocratic reversed-phase HPLC was employed for quantitative analysis using tetra-n-butylammonium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service