Skip to Content
Merck
All Photos(1)

Key Documents

PHR1304

Supelco

Butylated hydroxyanisole

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2(3)-t-Butyl-4-hydroxyanisole, 2(3)-t-Butylhydroquinone monomethyl ether, BHA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H3(OCH3)OH
CAS Number:
Molecular Weight:
180.24
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. B1212000
traceable to USP 1083008

vapor density

6.2 (vs air)

API family

butylhydroxyanisole

CofA

current certificate can be downloaded

autoignition temp.

599 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

58-60 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

O(C)c1cc(c(cc1)O)C(C)(C)C

InChI

1S/C11H16O2/c1-11(2,3)9-7-8(13-4)5-6-10(9)12/h5-7,12H,1-4H3

InChI key

MRBKEAMVRSLQPH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Butylated hydroxyanisole (BHA) may be used as a reference standard for the determination of the analyte in pharmaceutical creams[1] by spectrophotometry.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB3751 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

241.9 °F - Pensky-Martens closed cup

Flash Point(C)

116.6 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of miconazole in pharmaceutical creams using internal standard and second derivative spectrophotometry.
Wrobel K, et al.
Journal of Pharmaceutical and Biomedical Analysis, 20(1-2), 99-105 (1999)
[Modification of the processes of chemical mutagenesis and carcinogenesis by synthetic phenol antioxidants such as butylated hydroxyanisole].
K Demkowicz-Dobrzański et al.
Postepy biochemii, 38(1), 37-42 (1992-01-01)
In vivo studies on butylated hydroxyanisole.
F Iverson
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 37(9-10), 993-997 (1999-10-29)
H C Grice
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 26(8), 717-723 (1988-08-01)
Butylated hydroxyanisole (BHA) induces tumours of the squamous epithelium of the forestomach of rodents, but not at other sites. Although humans do not have squamous epithelium in their stomach the likelihood that BHA will induce tumours of the squamous epithelium
H Verhagen et al.
Chemico-biological interactions, 80(2), 109-134 (1991-01-01)
Butylated hydroxyanisole (BHA) is a synthetic food antioxidant used to prevent oils, fats and shortenings from oxidative deterioration and rancidity. This review depicts the current knowledge on BHA. The physical and chemical characteristics of BHA are summarized and its function

Articles

HPLC-UV method detects 9 antioxidants like propyl gallate in food using a Discovery®

HPLC-UV method detects 9 antioxidants like propyl gallate in food using a Discovery®

HPLC-UV method detects 9 antioxidants like propyl gallate in food using a Discovery®

HPLC-UV method detects 9 antioxidants like propyl gallate in food using a Discovery®

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service