Skip to Content
Merck
All Photos(1)

Key Documents

PHR1117

Supelco

Butylated Hydroxytoluene

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, Butylhydroxytoluene, DBPC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H2(CH3)OH
CAS Number:
Molecular Weight:
220.35
Beilstein:
1911640
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E321
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. B1215000
traceable to USP 1092708

vapor density

7.6 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

API family

butylated hydroxytoluene

CofA

current certificate can be downloaded

autoignition temp.

878 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

265 °C (lit.)

mp

69-73 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3

InChI key

NLZUEZXRPGMBCV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Butylated hydroxytoluene (BHT) is a phenolic antioxidant used as a food additive. It is frequently added to dried cereals, cooking oils and animal foods. BHT is also known to enhance the stability of pharmaceuticals, fat-soluble vitamins and cosmetics .

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB3695 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

260.6 °F - open cup

Flash Point(C)

127 °C - open cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactivity of butylated hydroxytoluene.
Lambert CR, et al.
Free Radical Biology & Medicine, 21(3), 395-400 (1996)
Butylated hydroxytoluene (BHT): a review.
H Babich
Environmental research, 29(1), 1-29 (1982-10-01)
Rio P Juni et al.
JACC. Basic to translational science, 4(5), 575-591 (2019-11-27)
The positive findings of the EMPA-REG OUTCOME trial (Randomized, Placebo-Controlled Cardiovascular Outcome Trial of Empagliflozin) on heart failure (HF) outcome in patients with type 2 diabetes mellitus suggest a direct effect of empagliflozin on the heart. These patients frequently have
Deliang Guo et al.
Science signaling, 2(101), ra82-ra82 (2009-12-17)
Glioblastoma, the most common malignant brain tumor, is among the most lethal and difficult cancers to treat. Although epidermal growth factor receptor (EGFR) mutations are frequent in glioblastoma, their clinical relevance is poorly understood. Studies of tumors from patients treated
A M Malkinson
Environmental mutagenesis, 5(3), 353-362 (1983-01-01)
Although the average American's daily consumption of BHT can be measured in milligrams, there are numerous reports that BHT causes organ damage in laboratory animals. Only a few genotoxic effects of BHT have been reported, however, including mutagenicity in the

Articles

HPLC-UV method detects 9 antioxidants like propyl gallate in food using a Discovery®

HPLC-UV method detects 9 antioxidants like propyl gallate in food using a Discovery®

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service