Skip to Content
Merck
All Photos(1)

Key Documents

20021

Sigma-Aldrich

3-tert-Butyl-4-hydroxyanisole

≥98% (sum of isomers, GC), ≤10% 2-BHA basis (GC)

Synonym(s):

2-tert-Butyl-4-methoxyphenol, 3-BHA, BHA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16O2
CAS Number:
Molecular Weight:
180.24
Beilstein:
1867499
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98% (sum of isomers, GC)

form

solid

composition

2-BHA, ≤10% GC
3-BHA, ≥90% GC

impurities

≤1% 4-hydroxyanisole

ign. residue

≤0.05%

mp

48-63 °C

solubility

ethanol: soluble 1 g/10 mL, clear, colorless to faint yellow or tan

SMILES string

COc1ccc(O)c(c1)C(C)(C)C

InChI

1S/C11H16O2/c1-11(2,3)9-7-8(13-4)5-6-10(9)12/h5-7,12H,1-4H3

InChI key

MRBKEAMVRSLQPH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The voltammetric behavior of 3-tert-butyl-4-hydroxyanisole at a polymer electrode modified with nickel phthalocyanine (as electron mediator) was studied.

Application

  • Regulation of Smad signaling in mesenchymal stem cells: 3-tert-Butyl-4-hydroxyanisole disrupts the differentiation of C3H10T1/2 mesenchymal stem cells into brown adipocytes by modulating Smad signaling pathways, with potential implications for obesity and metabolic syndrome research (Wang et al., 2023).

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C de la Fuente et al.
Talanta, 49(2), 441-452 (2008-10-31)
The voltammetric behaviour of the antioxidants 3-tert-butyl-4-hydroxyanisole (BHA) and tert-butylhydroquinone (TBHQ), at a polymer electrode modified with nickel phthalocyanine as electron mediator, is described, and an electroanalytical method for the determination of these antioxidants based on their electrochemical oxidation on
K Tajima et al.
Drug metabolism and disposition: the biological fate of chemicals, 20(6), 816-820 (1992-11-01)
3-Tert-butyl-4-hydroxyanisole (3-BHA) was metabolized in the presence of horseradish peroxidase and hydrogen peroxide to 2-tert-butyl-p-benzoquinone (TBQ), 2,3-epoxy-5-tert-butyl-1,4-benzoquinone (TBQ-epoxide), and two known dimers. The formation of TBQ from 3-BHA required both horseradish peroxidase and hydrogen peroxide. When 2.5 mM 3-BHA was
A Matsuoka et al.
Mutation research, 241(2), 125-132 (1990-06-01)
The mutagenicity of 3-tert-butyl-4-hydroxyanisole (BHA) and its metabolites was investigated in the reverse mutation assay using S. typhimurium strains and the chromosomal aberration test in vitro using a Chinese hamster fibroblast cell line, CHL. BHA, tert-butylhydroquinone (BHQ), tert-butylquinone (BQ) and
Mengxiao Xie et al.
Journal of autoimmunity, 102, 96-113 (2019-05-28)
The subset of regulatory T (Treg) cells, with its specific transcription Foxp3, is a unique cell type for the maintenance of immune homeostasis by controlling effector T (Teff) cell responses. Although it is common that a defect in Treg cells
L K Lam et al.
Carcinogenesis, 12(12), 2311-2315 (1991-12-01)
Carcinogen-induced aberrant crypts (AC) of the colon are a precancerous state that leads to malignancy. The inhibition of AC formation by chemopreventive agents was evaluated in this study. Colon AC were induced by 1,2-dimethylhydrazine (DMH) in 3 weeks in CF1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service