Skip to Content
Merck
All Photos(1)

Key Documents

P2700000

Prednisolone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,4-Pregnadiene-11β,17α,21-triol-3,20-dione, 1-Dehydrocortisol, 1-Dehydrohydrocortisone, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O5
CAS Number:
Molecular Weight:
360.44
Beilstein:
1354103
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

prednisolone

manufacturer/tradename

EDQM

mp

240 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O=C1C=C[C@@]2(C)C(CC[C@]3([H])[C@]2([H])[C@@H](O)C[C@@]4(C)[C@@]3([H])CC[C@]4(O)C(CO)=O)=C1

InChI

1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-16,18,22,24,26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1

InChI key

OIGNJSKKLXVSLS-VWUMJDOOSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Prednisolone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vappu Rantalaiho et al.
Annals of the rheumatic diseases, 73(11), 1954-1961 (2013-08-03)
To study whether adding initial infliximab to remission-targeted initial combination-DMARD treatment improves the long-term outcomes in patients with early rheumatoid arthritis (RA). Ninety-nine patients with early, DMARD-naïve RA were treated with a triple combination of DMARDs, starting with methotrexate (max
Axel Heidenreich et al.
European journal of cancer (Oxford, England : 1990), 50(6), 1090-1099 (2014-02-04)
Cabazitaxel/prednisone has been shown to prolong survival versus mitoxantrone/prednisone in patients with metastatic castration-resistant prostate cancer (mCRPC) that has progressed during or after docetaxel. Subsequently, compassionate-use programmes (CUPs) and expanded-access programmes (EAPs) were established worldwide, allowing access to cabazitaxel before
J A P Spijkers-Hagelstein et al.
Leukemia, 28(4), 761-769 (2013-08-21)
Successful treatment results for MLL-rearranged Acute Lymphoblastic Leukemia (ALL) in infants remain difficult to achieve. Significantly contributing to therapy failure is poor response to glucocorticoids (GCs), like prednisone. Thus, overcoming resistance to these drugs may be a crucial step towards
Eng Cern Gan et al.
International forum of allergy & rhinology, 4(11), 877-884 (2014-08-20)
The endoscopically magnified operative field in functional endoscopic sinus surgery (FESS) makes even a small amount of bleeding a potentially significant hindrance. It is thought that irrigation with hot saline during surgery may improve surgical field of view by producing
Alexander W Hauswirth et al.
Leukemia research, 28(3), 249-257 (2003-12-23)
Aggressive systemic mastocytosis (ASM) is a hematopoietic neoplasm characterized by infiltration of visceral organs by neoplastic mast cells (MCs) with consecutive organopathy and respective clinical and laboratory findings (so called C-Findings). Whereas, it is generally appreciated that patients with ASM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service