Skip to Content
Merck
All Photos(1)

Key Documents

H1300000

Hydrocortisone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

11β,17α,21-Trihydroxypregn-4-ene-3,20-dione, 17-Hydroxycorticosterone, 4-Pregnene-11β,17α,21-triol-3,20-dione, Cortisol, Kendall’s compound F, Reichstein’s substance M

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O5
CAS Number:
Molecular Weight:
362.46
Beilstein:
1354819
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

hydrocortisone

manufacturer/tradename

EDQM

mp

211-214 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)CO

InChI

1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1

InChI key

JYGXADMDTFJGBT-VWUMJDOOSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hydrocortisone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Primary glucocorticoid secreted by the adrenal cortex.
Primary glucocorticoid secreted by the adrenal cortex. It has three times the anti-inflammatory potency of corticosterone but much lower Na2+ retention potency.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorenz Goette et al.
Psychoneuroendocrinology, 54, 115-123 (2015-02-24)
Social competition is a fundamental mechanism of evolution and plays a central role in structuring individual interactions and communities. Little is known about the factors that affect individuals' competitive success, particularly in humans. Key factors might include stress, a major
Florian Hardeveld et al.
Psychoneuroendocrinology, 50, 62-71 (2014-09-03)
Although HPA-axis activity has been studied extensively in relation to depression, there is no consensus whether HPA-axis parameters predicts major depressive disorder (MDD) recurrence. We investigated whether HPA-axis parameters (cortisol awakening response (CAR), the dexamethasone suppression test (DST) and evening
Rachel Yehuda et al.
The American journal of psychiatry, 171(8), 872-880 (2014-05-17)
Differential effects of maternal and paternal posttraumatic stress disorder (PTSD) have been observed in adult offspring of Holocaust survivors in both glucocorticoid receptor sensitivity and vulnerability to psychiatric disorder. The authors examined the relative influences of maternal and paternal PTSD
Lex W Doyle et al.
Neonatology, 98(2), 111-117 (2010-02-13)
Corticosteroids have been used widely after birth in preterm infants with respiratory failure; hydrocortisone may be preferable to other corticosteroids for this purpose. To determine if postnatal hydrocortisone is useful to prevent or treat bronchopulmonary dysplasia (BPD) in preterm infants.
Marianne Oksnes et al.
The Journal of clinical endocrinology and metabolism, 99(5), 1665-1674 (2014-02-13)
Conventional glucocorticoid replacement therapy fails to mimic the physiological cortisol rhythm, which may have implications for morbidity and mortality in patients with Addison's disease. The objective of the study was to compare the effects of continuous sc hydrocortisone infusion (CSHI)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service