Skip to Content
Merck
All Photos(1)

Documents

1555005

USP

Prednisolone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1,4-Pregnadiene-11β,17α,21-triol-3,20-dione, 1-Dehydrocortisol, 1-Dehydrohydrocortisone, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O5
CAS Number:
Molecular Weight:
360.44
Beilstein:
1354103
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

prednisolone

manufacturer/tradename

USP

mp

240 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1C=C[C@@]2(C)C(CC[C@]3([H])[C@]2([H])[C@@H](O)C[C@@]4(C)[C@@]3([H])CC[C@]4(O)C(CO)=O)=C1

InChI

1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-16,18,22,24,26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1

InChI key

OIGNJSKKLXVSLS-VWUMJDOOSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.

Application

It is intended to be used as a USP reference standard in specified quality tests and assays.
Also used to prepare standard and system suitability solutions as described in the following United States Pharmacopeia (USP) monographs:
  • Prednisolone
  • Prednisolone Compounded Oral Suspension, Veterinary
  • Prednisolone Sodium Phosphate
  • Dexamethasone Oral Solution
  • Tetracycline Hydrochloride, Novobiocin Sodium, and Prednisolone Tablets
  • Prednisolone Oral Solution
  • Prednisolone Tablets
  • Prednisolone Sodium Phosphate Injection
  • Prednisolone Sodium Phosphate Ophthalmic Solution
  • Hydrocortisone Gel

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tetracycline Hydrochloride, Novobiocin Sodium, and Prednisolone Tablets
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 4312-4312 (2013)
Prednisolone Sodium Phosphate Ophthalmic Solution
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 3668-3668 (2020)
Dexamethasone Oral Solution
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(6), 1288-1288 (2014)
Prednisolone
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 30(5), 3659-3659 (2020)
Prednisolone Oral Solution
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 28(2), 3661-3661 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service