Skip to Content
Merck
All Photos(1)

Documents

M1850000

Metronidazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Methyl-5-nitroimidazole-1-ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O3
CAS Number:
Molecular Weight:
171.15
Beilstein:
611683
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

metronidazole

manufacturer/tradename

EDQM

mp

159-161 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1=NC=C([N+]([O-])=O)N1CCO

InChI

1S/C6H9N3O3/c1-5-7-4-6(9(11)12)8(5)2-3-10/h4,10H,2-3H2,1H3

InChI key

VAOCPAMSLUNLGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Metronidazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonja Löfmark et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 50 Suppl 1, S16-S23 (2010-01-14)
Metronidazole has been used for the treatment of infections for >45 years and is still successfully used for the treatment of trichomoniasis, amoebiasis, and giardiasis. Anaerobic bacterial infections caused by Bacteroides species, fusobacteria, and clostridia respond favorably to metronidazole therapy.
Fabrizio Sgolastra et al.
Journal of periodontology, 83(10), 1257-1269 (2012-01-10)
The combination of Amoxicillin and metronidazole (AMX/MET) as an adjunctive treatment to scaling root planing (SRP) has been proposed for the treatment of chronic periodontitis; however, its effectiveness and clinical safety remain to be defined. The purpose of the present
Dina Zandbergen et al.
Journal of periodontology, 84(3), 332-351 (2012-05-23)
The treatment of periodontitis frequently begins with a non-surgical phase that includes scaling and root planing (SRP) and, on occasion, the use of systemic antibiotics. The goal of this review is to systematically evaluate the data concerning the effect of
Michael John Aldape et al.
The Journal of antimicrobial chemotherapy, 70(1), 153-159 (2014-08-26)
Clostridium difficile infection (CDI) is mediated by potent extracellular toxins and is spread largely via bacterial spores. We and others have shown that some antibiotics stimulate C. difficile toxin production in a strain-specific manner; however, the effects of newer anti-C.
Fabrizio Sgolastra et al.
Journal of periodontology, 83(6), 731-743 (2011-11-05)
The systemic use of combined amoxicillin and metronidazole (AMX/MET) as an adjunctive treatment to full-mouth scaling and root planing (FMSRP) has been proposed for the treatment of generalized aggressive periodontitis; however, its effectiveness and clinical safety remain to be defined.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service