Skip to Content
Merck
All Photos(1)

Key Documents

H1425000

Hydrocortisone hydrogen succinate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Hydrocortisone 21-hemisuccinate, 11β,17α,21-Trihydroxy-4-pregnene-3,20-dione 21-hemisuccinate, Cortisol 21-hemisuccinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H34O8
CAS Number:
Molecular Weight:
462.53
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

hydrocortisone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)COC(=O)CCC(O)=O)[C@@]4(C)C[C@H](O)[C@H]23

InChI

1S/C25H34O8/c1-23-9-7-15(26)11-14(23)3-4-16-17-8-10-25(32,24(17,2)12-18(27)22(16)23)19(28)13-33-21(31)6-5-20(29)30/h11,16-18,22,27,32H,3-10,12-13H2,1-2H3,(H,29,30)/t16-,17-,18-,22+,23-,24-,25-/m0/s1

InChI key

VWQWXZAWFPZJDA-CGVGKPPMSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hydrocortisone hydrogen succinate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaolin Qi et al.
American journal of ophthalmology, 158(6), 1192-1198 (2014-08-20)
To investigate the morphologic changes and outcomes of the amniotic membrane as a carrier for allogeneic cultivated limbal epithelial transplantation. Prospective, noncomparative, interventional study. A total of 16 eyes receiving allogeneic cultivated limbal epithelial transplantation with amniotic membrane as a
Lucia Gagliardi et al.
The Journal of clinical endocrinology and metabolism, 99(11), 4149-4157 (2014-08-16)
Patients with Addison's disease (AD) report impaired subjective health status (SHS). Since cortisol exhibits a robust circadian cycle that entrains other biological clocks, impaired SHS may be due to the noncircadian cortisol profile achieved with conventional glucocorticoid replacement. Continuous subcutaneous
Maureen Keller-Wood et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 307(4), R405-R413 (2014-06-13)
In normal pregnancy, cortisol increases; however, further pathological increases in cortisol are associated with maternal and fetal morbidities. These experiments were designed to test the hypothesis that increased maternal cortisol would increase maternal glucose concentrations, suppress fetal growth, and impair
Takeshi Soma et al.
PloS one, 9(10), e110987-e110987 (2014-10-25)
We assessed the maintenance and distribution of epithelial stem/progenitor cells after corneal reconstruction using tissue-engineered oral mucosal cell sheets in a rat model. Oral mucosal biopsy specimens were excised from green fluorescent protein (GFP) rats and enzymatically treated with Dispase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service