Skip to Content
Merck
All Photos(1)

Key Documents

B8522

Sigma-Aldrich

Brilliant Blue G solution

Concentrate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

liquid

technique(s)

microbe id | staining: suitable

color

dark blue

suitability

suitable for (Suitable for for staining protein in polyacrylamide and agarose gels)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[Na+].CCOc1ccc(Nc2ccc(cc2)C(\c3ccc(cc3C)N(CC)Cc4cccc(c4)S([O-])(=O)=O)=C5\C=CC(\C=C5C)=[N+](\CC)Cc6cccc(c6)S([O-])(=O)=O)cc1

InChI

1S/C47H49N3O7S2.Na/c1-6-49(31-35-11-9-13-43(29-35)58(51,52)53)40-21-25-45(33(4)27-40)47(37-15-17-38(18-16-37)48-39-19-23-42(24-20-39)57-8-3)46-26-22-41(28-34(46)5)50(7-2)32-36-12-10-14-44(30-36)59(54,55)56;/h9-30H,6-8,31-32H2,1-5H3,(H2,51,52,53,54,55,56);/q;+1/p-1

InChI key

RWVGQQGBQSJDQV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Brilliant Blue G is a methanol based stain which is especially designed for staining protein in polyacrylamide gels, but will also stain protein in agarose gels (IEF). The stain contains methanol and acetic acid so gels do not require fixing prior to staining. Water is added directly to the bottle to produce 1 liter of ready-to-use staining solution.

Application

Brilliant Blue G solution has been used for protein band detection after SDS-PAGE (sodium dodecyl sulfate polyacrylamide gel electrophoresis). It is also used for the protein staining of cells on microscopic slide.

Quantity

One bottle equals one liter working solution. 300 ml of brilliant blue G solution concentrate is supplied in the bottle.

Reconstitution

After dilution with deionized water to a final volume of one liter, solution will contain 0.1% (w/v) Brilliant Blue G, 25% (v/v) methanol and 5% (v/v) acetic acid.

Analysis Note

This product has been tested for suitability on SDS-PAGE.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emerin suppresses Notch signaling by restricting the Notch intracellular domain to the nuclear membrane.
Biochimica et Biophysica Acta, 1864, 303-303 (2017)
Suma Tiruvayipati et al.
PloS one, 15(3), e0228871-e0228871 (2020-03-03)
Exosomes are extracellular vesicles (EVs) of ~20-200 nm diameter that shuttle DNAs, RNAs, proteins and other biomolecules between cells. The large number of biomolecules present in exosomes demands the frequent use of high-throughput analysis. This, in turn, requires technical replicates
Effect of an experimental proteasome inhibitor on the cytoskeleton, cytosolic protein turnover, and induction in the neuronal cells in vitro.
Csizmadia V, et al.
Neurotoxicology, 29, 232-232 (2008)
Ahmed H
Principles and Reactions of Protein Extraction, Purification, and Characterization (2004)
Starlee Lively et al.
Frontiers in cellular neuroscience, 12, 115-115 (2018-05-22)
The cytokine, transforming growth factor β1 (TGFβ1), is up-regulated after central nervous system (CNS) injuries or diseases involving microglial activation, and it has been proposed as a therapeutic agent for treating neuroinflammation. Microglia can produce and respond to TGFβ1. While

Articles

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service