Skip to Content
Merck
All Photos(2)

Key Documents

A4628

Sigma-Aldrich

Bovine Serum Albumin solution

5% in 0.85% sodium chloride, aseptically filled

Synonym(s):

Albumin solution from bovine serum, BSA solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27
Pricing and availability is not currently available.

biological source

bovine

Quality Level

sterility

aseptically filled

Assay

4.8-5.5% protein basis (biuret)

form

solution

purified by

heat shock fractionation

does not contain

stabilizer or preservative

origin

USA origin

concentration

4.8-5.5% (biuret)
5% in 0.85% sodium chloride

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

General description

Bovine serum albumin, a large molecular weight protein[1], accounts for around 10% of total whey proteins.[2] It is a source of essential amino acids and is the third most abundant whey protein found in milk.[2] BSA is biodegradable, biocompatible, nontoxic, and nonimmunogenic.[3]

Application

Bovine Serum Albumin solution has been used:
  • as a component in Opti-minimum essential medium (MEM) to serve as the transfection media to study the effect of albumin on transfection efficiency[3]
  • as a component in 0.1% Tween-20/PBS (PBST) in diluting primary antibodies to incubate cells[3]
  • to incubate hydroxyapatite (HA) for in vitro study[4]

Biochem/physiol Actions

Bovine serum albumin is capable of binding free fatty acids, other lipids, and flavor compounds. Because of its low cost, tolerance, and ease of preparation, BSA has been widely used in drug delivery. BSA can be used as a colloidal delivery system. It is also appealing as a nanoparticle matrix due to its low cost, ease of purification, and aqueous solubility. BSA nanoparticles can load several drugs and bioactive compounds due to drug binding sites in the BSA molecule.[3]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Daculsi et al.
Journal of biomedical materials research, 47(2), 228-233 (1999-08-17)
The role of adhesion molecules like osteopontin and bone sialoprotein, both containing the Arg-Gly-Asp sequence have been shown to have a role in mineral formation, whereas fibronectin (FN), another adhesive protein, was never studied during the mineralization processes. The formation
Heterogeneity, Fractionation, and Isolation
12th International scientific conference : Achievements in mechanical and materials engineering. (2022)
An overview of biopolymer nanostructures for encapsulation of food ingredients
3D Printing and Biofabrication, 1-35 (2019)
Whey proteins in functional foods
3D Printing and Biofabrication, 637-663 (2019)
Hong Yao et al.
Biomaterials, 30(29), 5793-5803 (2009-07-21)
The success of gene therapy relies on a safe and effective gene delivery system. In this communication, we describe the use of folate grafted PEI(600)-CyD (H(1)) as an effective polyplex-forming plasmid delivery agent with low toxicity. The structures of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service