Skip to Content
Merck
All Photos(1)

Key Documents

A0225000

Acitretin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O3
CAS Number:
Molecular Weight:
326.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

acitretin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(C)c(\C=C\C(C)=C\C=C\C(C)=C\C(O)=O)c(C)c1C

InChI

1S/C21H26O3/c1-14(8-7-9-15(2)12-21(22)23)10-11-19-16(3)13-20(24-6)18(5)17(19)4/h7-13H,1-6H3,(H,22,23)/b9-7+,11-10+,14-8+,15-12+

InChI key

IHUNBGSDBOWDMA-AQFIFDHZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Acitretin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Synthetic retinoid that is a metabolite of etretinate. Preferentially binds to cellular retinoic acid binding proteins (CRABPs).

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justin Cheeley et al.
Journal of the American Academy of Dermatology, 68(2), 247-254 (2012-08-25)
Bexarotene is the only Food and Drug Administration-approved retinoid for the treatment of cutaneous T-cell lymphoma (CTCL) and is associated with a relatively high frequency of adverse effects. Acitretin has anecdotally been reported to be effective for CTCL. We sought
Vincenzo Bettoli et al.
The Journal of dermatological treatment, 24(3), 235-237 (2012-11-15)
The chemoprevention refers to the use of various types of chemical agents for preventing carcinogenic progression. Systemic retinoids are the most studied chemopreventive agents due to their capacity to regulate cell proliferation and their demonstrated efficacy in several clinical studies.
Sandra Philipp et al.
Journal der Deutschen Dermatologischen Gesellschaft = Journal of the German Society of Dermatology : JDDG, 10(11), 821-837 (2012-09-01)
Monotherapy with TNF-α inhibitors does not always produce a sufficient response in psoriasis patients. Combinations of TNF-α antagonists such as adalimumab with systemic antipsoriatic therapies such as methotrexate are not approved for use in psoriasis, and the published data are
Systemic retinoid therapy for chemoprevention of nonmelanoma skin cancer in a patient treated with vemurafenib.
Rachael Anforth et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 30(19), e165-e167 (2012-06-06)
Howa Yeung et al.
Journal of the American Academy of Dermatology, 68(1), 64-72 (2012-08-01)
Despite widespread dissatisfaction and low treatment persistence in moderate to severe psoriasis, patients' reasons behind treatment discontinuation remain poorly understood. We sought to characterize patient-reported reasons for discontinuing commonly used treatments for moderate to severe psoriasis in real-world clinical practice.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service