Skip to Content
Merck

Skip To

44707

Acitretin

≥98.0% (HPLC)

Synonym(s):

9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H26O3
CAS Number:
Molecular Weight:
326.43
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
259-474-4
MDL number:
Assay:
≥98.0% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (HPLC)

form

powder

SMILES string

COc1cc(C)c(\C=C\C(C)=C\C=C\C(C)=C\C(O)=O)c(C)c1C

InChI

1S/C21H26O3/c1-14(8-7-9-15(2)12-21(22)23)10-11-19-16(3)13-20(24-6)18(5)17(19)4/h7-13H,1-6H3,(H,22,23)/b9-7+,11-10+,14-8+,15-12+

InChI key

IHUNBGSDBOWDMA-AQFIFDHZSA-N

Application

Acitretin, a retinoid that binds to nuclear receptors and regulates gene expression, is a potent activator of the α-secretase ADAM10 gene expression and apoptosis inducer via the CD95 signalling pathway. Acitretin is a systemic retinoid drug used in the treatment of severe psoriasis.

Biochem/physiol Actions

Synthetic retinoid that is a metabolite of etretinate. Preferentially binds to cellular retinoic acid binding proteins (CRABPs).


Still not finding the right product?

Explore all of our products under Acitretin


signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

wgk

WGK 3

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters