Skip to Content
Merck
All Photos(1)

Documents

1011018

USP

Acitretin Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(2Z,4E,6E,8E)-9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid, 13-cis-Acitretin, Isoacitretin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O3
CAS Number:
Molecular Weight:
326.43
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

acitretin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

InChI

1S/C21H26O3/c1-14(8-7-9-15(2)12-21(22)23)10-11-19-16(3)13-20(24-6)18(5)17(19)4/h7-13H,1-6H3,(H,22,23)/b9-7+,11-10+,14-8+,15-12-

InChI key

IHUNBGSDBOWDMA-UGOGCBOOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Acitretin Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Acitretin

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D J Lospinoso et al.
Lupus, 22(8), 851-854 (2013-06-14)
Lupus erythematosus/lichen planus overlap syndrome is a rare disorder combining the clinical, histological and immunopathological features of both lupus erythematosus (LE) and lichen planus (LP). Cutaneous lesions mostly affect the distal arms, legs, face and trunk. Palmoplantar involvement is felt
Matthias Lahner et al.
Technology and health care : official journal of the European Society for Engineering and Medicine, 21(3), 265-269 (2013-06-26)
Adhesive capsulitis (AC) is characterized by a limited active and passive motion. Although the exact pathology remains unknown, a number of contributing factors are discussed. AC has probably been caused by the Re-PUVA therapy (PUVA irradiation plus acitretin) of a
Evren Odyakmaz Demirsoy et al.
Journal of drugs in dermatology : JDD, 12(9), 1039-1043 (2013-09-05)
Nails, one of the most visible sites of body, are frequently involved in psoriasis and accepted as the most difficult site for topical treatment because of their anatomical structure. Healing of the psoriatic nails usually occurs when systemic therapy is
Somenath Sarkar et al.
Indian journal of pharmacology, 45(1), 89-90 (2013-04-02)
Pseudotumor cerebri (PTC) is a rare neurological disorder characterized by increased intracranial pressure in absence of any intra-cranial space-occupying lesion. It is mostly due to impairment of drainage of CSF from arachnoid villi. Clinically pseudotumor cerebri presents with headache, diplopia
Liraglutide in combination with acitretin for severe recalcitrant psoriasis.
C T Reid et al.
The British journal of dermatology, 169(1), 230-231 (2013-07-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service