Skip to Content
Merck
All Photos(3)

Documents

933864

Sigma-Aldrich

Dimethyl isosorbide

greener alternative

BioRenewable, anhydrous, DMF and NMP substitute, ≥99%

Synonym(s):

Isosorbide dimethyl ether, 1,4:3,6-Dianhydrosorbitol 2,5-dimethyl ether, 1,4:3,6-Dianhydro-2,5-di-O-methyl-D-glucitol, 2,5-Di-O-methyl-1,4:3,6-dianhydro-D-glucitol, DMI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O4
CAS Number:
Molecular Weight:
174.19
Beilstein:
80854
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.21

Quality Level

Assay

≥99%

form

liquid

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

colorless

refractive index

n20/D 1.461 (lit.)

bp

93-95 °C/0.1 mmHg (lit.)

density

1.15 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

CO[C@H]1COC2[C@@H](COC12)OC

InChI

1S/C8H14O4/c1-9-5-3-11-8-6(10-2)4-12-7(5)8/h5-8H,3-4H2,1-2H3/t5-,6+,7-,8-/m1/s1

InChI key

MEJYDZQQVZJMPP-ULAWRXDQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are dedicated to providing Greener Alternative Products that adhere to The 12 Principles of Greener Chemistry. Dimethyl isosorbide (DMI) is a bio-based solvent that aligns with the principles of "Use of Renewable Feedstocks" and "Safer Solvents and Auxiliaries", making it a sustainable alternative to dipolar aprotic solvents like NMP and DMF. DMI′s high solubilization capacity makes it suitable to be used as a co-solvent for nonpolar drugs and in organic synthesis. Moreover, its exceptional properties, such as non-toxicity, non-irritation, and biodegradability, make it a safe and eco-friendly choice for both customers and the environment.

Application

  • Polymer membranes preparation
  • Peptide synthesis
  • Cross-coupling reactions
  • Co-solvent to improve solubility of nonpolar compounds
  • Lithium battery recycling

Features and Benefits

  • Bio-based
  • Made from Renewable Resource - straw
  • Non-toxic solvent
  • Non-irritation
  • Biodegradability
  • High solubility potential

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dimethyl Isosorbide As a Green Solvent for Sustainable Ultrafiltration and Microfiltration Membrane Preparation
Russo F, et al.
ACS sustainable chemistry & engineering, 8, 659?668-659?668 (2020)
Nickel-Catalyzed Reductive Cross-Coupling of Aryl Bromides with Vinyl Acetate in Dimethyl Isosorbide as a Sustainable Solvent
Mincong S, et al.
Organic Letters, 24, 354?358-354?358 (2022)
Dimethylisosorbide (DMI) as a Bio-Derived Solvent for Pd-Catalyzed Cross-Coupling Reactions
Wilson K L, et al.
Synlett, 29, 2293-2297 (2018)
A sustainable approach to cathode delamination using a green solvent
Onurcan Buken, Kayla Mancinia and Amrita Sarkar
Royal Society of Chemistry Advances, 11, 27356-27368 (2021)
Greening Fmoc/tBu solid-phase peptide synthesis
Musaimi O A, et al.
Green Chemistry, 4, 996-1018 (2020)

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service