Skip to Content
Merck
All Photos(1)

Documents

V403

Sigma-Aldrich

γ-Valerolactone

ReagentPlus®, 99%

Synonym(s):

γ-Valerolactone, gamma-Valerolactone, γ-Methyl-γ-butyrolactone, 4,5-Dihydro-5-methyl-2(3H)-furanone, 4-Hydroxypentanoic acid lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O2
CAS Number:
Molecular Weight:
100.12
Beilstein:
80420
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.45 (vs air)

product line

ReagentPlus®

Assay

99%

refractive index

n20/D 1.432 (lit.)

bp

207-208 °C (lit.)
82-85 °C/10 mmHg (lit.)

mp

−31 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

SMILES string

CC1CCC(=O)O1

InChI

1S/C5H8O2/c1-4-2-3-5(6)7-4/h4H,2-3H2,1H3

InChI key

GAEKPEKOJKCEMS-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

γ-valerolactone (GVL) is found naturally in fruits and often used as a fuel additive and food ingredient. It is prominently employed in the production of both energy and carbon-based products.

Application

γ-valerolactone (GVL) can be used as a green solvent:
  • To transform lignocellulose into furfural using a solid acid catalyst, H-mordenite.
  • To synthesize phosphatidylserine.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

γ-Valerolactone-a sustainable liquid for energy and carbon-based chemicals
Horvath I, et al.
Green Chemistry, 10(2), 238-242 (2008)
Highly efficient synthesis of phosphatidylserine in the eco-friendly solvent γ-valerolactone
Duan Z-Q and Hu Fei
Green Chemistry, 14(6), 1581-1583 (2012)
Conversion of levulinic acid and formic acid into γ-valerolactone over heterogeneous catalysts.
Li Deng et al.
ChemSusChem, 3(10), 1172-1175 (2010-09-28)
Catalytic conversion of biomass-derived carbohydrates into gamma-valerolactone without using an external H2 supply.
Li Deng et al.
Angewandte Chemie (International ed. in English), 48(35), 6529-6532 (2009-07-25)
Santiago Aparicio et al.
Physical chemistry chemical physics : PCCP, 11(30), 6455-6467 (2009-10-08)
In this work, an experimental and computational study on the properties and molecular-level liquid structure of gamma-butyrolactone and gamma-valerolactone is reported. These fluids are selected because of their possible use as alternative green solvents considering their favorable environmental and toxicological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service