Skip to Content
Merck
All Photos(3)

Key Documents

40727

Sigma-Aldrich

1,3-Dimethyl-2-imidazolidinone

≥99.0% (GC)

Synonym(s):

N,N′-Dimethylethyleneurea, DMEU, DMI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10N2O
CAS Number:
Molecular Weight:
114.15
Beilstein:
108808
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (GC)

form

liquid

refractive index

n20/D 1.472 (lit.)

bp

224-226 °C (lit.)

solubility

toluene: soluble(lit.)
water: miscible

density

1.056 g/mL at 25 °C (lit.)

SMILES string

CN1CCN(C)C1=O

InChI

1S/C5H10N2O/c1-6-3-4-7(2)5(6)8/h3-4H2,1-2H3

InChI key

CYSGHNMQYZDMIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Dimethyl-2-imidazolidinone (DMI) is an imidazolidine derivative. It is reported to act as a promoter by minimizing the formation of dialkylation byproducts and accelerating the rate of monoalkylation of γ-butyrolactone. Conversion of CO2 to form lower alcohols by homogeneous catalytic hydrogenation using DMI as solvent has been investigated.

Application

1,3-Dimethyl-2-imidazolidinone may be used:
  • As substitute solvent for hexamethylphosphoric triamide (HMPA) in the synthesis of 1,2-bis(trimethylsilyl)benzene.
  • As solvent during the α-regioselective prenylation of imine.
  • As component of mobile phase for the size-exclusion chromatographic analysis of cellulose.

Other Notes

Solvent used in various synthetic organic transformations. Studied in the formation of functionally stabilized hydrosilanediyl-transition metal complexes produced photochemically from arylsilanes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Case Study of a γ-Butyrolactone Alkylation with 1,3-Dimethyl-2-imidazolidinone as a Promoter.
Li B, et al.
Organic Process Research & Development, 5(6), 609-611 (2001)
Tsugio Kitamura et al.
The Journal of organic chemistry, 78(7), 3421-3424 (2013-03-20)
A practical and safe synthesis of 1,2-bis(trimethylsilyl)benzene from 1,2-dichlorobenzene and Me3SiCl was achieved by use of a hybrid metal of Mg and CuCl in the presence of LiCl in 1,3-dimethyl-2-imidazolidinone (DMI). This method does not require a toxic HMPA, provides
Li-Ming Zhao et al.
Organic letters, 14(3), 886-889 (2012-01-24)
A highly α-regioselective prenylation of imines has been successfully developed. The efficiency of this approach is confirmed by a wide range of imines including N- and C-aryl aldimines, N-alkyl aldimines, C-alkyl aldimines, and N- and C-aryl ketimines. The approach uses
SEC-MALLS analysis of cellulose using LiCl/1, 3-dimethyl-2-imidazolidinone as an eluent.
Yanagisawa M, et al.
Cellulose, 11(2), 169-176 (2004)
Highly selective hydrogenation of CO2 into C2+ alcohols by homogeneous catalysis.
Qian Q, et al.
Chemical Science, 6(10), 5685-5689 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service