Skip to Content
Merck
All Photos(1)

Key Documents

90781

Sigma-Aldrich

Tributylamine

puriss. plus, ≥99.5% (GC)

Synonym(s):

Tri-n-butylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3N
CAS Number:
Molecular Weight:
185.35
Beilstein:
1698872
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
technique(s):
GC/GC: suitable
bp:
216 °C (lit.)
vapor pressure:
0.3 mmHg ( 20 °C)
2.4 mmHg ( 55 °C)

vapor density

6.38 (vs air)

Quality Level

vapor pressure

0.3 mmHg ( 20 °C)
2.4 mmHg ( 55 °C)

grade

puriss. plus

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

410 °F

expl. lim.

6 %

technique(s)

GC/GC: suitable

impurities

≤0.2% water

refractive index

n20/D 1.428 (lit.)
n20/D 1.429

bp

216 °C (lit.)

mp

−70 °C (lit.)

density

0.778 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCCCN(CCCC)CCCC

InChI

1S/C12H27N/c1-4-7-10-13(11-8-5-2)12-9-6-3/h4-12H2,1-3H3

InChI key

IMFACGCPASFAPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tributylamine is a tertiary amine. It has been reported to prevent the corrosion of mild steel in HCl solution.

Application

Tributylamine (n-Bu3N) may be used in the following studies:
  • To compose the mobile phase for the liquid chromatography tandem mass spectrometry (LC-MS/MS) analysis.
  • Preparation of PdCl,N and PdOAc,N (palladium nanoparticles).
  • As an additive during the synthesis of 3-bromothieno[3,2-c]pyridine-4-(5H)-one.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Palladium nanoparticles obtained from palladium salts and tributylamine in molten tetrabutylammonium bromide: their use for hydrogenolysis-free hydrogenation of olefins.
Le Bras J, et al.
New. J. Chem., 28(12), 1550-1553 (2004)
Tributylamine as corrosion inhibitor for mild steel in hydrochloric acid.
Bastidas JM, et al.
J. Mater. Sci., 35(11), 2637-2642 (2000)
Thermal Synthesis of 3-Bromothieno [3, 2-c] pyridin-4-(5H)-one: A Telescoped Procedure with Tributylamine.
Boros EE and Kaldor I.
Journal of Heterocyclic Chemistry, 52(1), 302-305 (2015)
Alexander S Misharin et al.
Analytical chemistry, 77(2), 459-470 (2005-01-15)
The analytical performance of an atmospheric pressure sampling, multiple-channel, high-throughput mass spectrometer was investigated using samples of a variety of types. The instrument, based on an array of cylindrical ion traps, was built with four independent channels and here is
D Sulzer et al.
Journal of neurochemistry, 60(2), 527-535 (1993-02-01)
Amphetamine-like psychostimulants are thought to produce rewarding effects by increasing dopamine levels at mesolimbic synapses. Paradoxically, dopamine uptake blockers, which generally increase extracellular dopamine, inhibit amphetamine-induced dopamine overflow. This effect could be due to either inhibition of amphetamine uptake or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service