Skip to Content
Merck
All Photos(1)

Key Documents

90780

Sigma-Aldrich

Tributylamine

puriss. p.a., ≥99.0% (GC)

Synonym(s):

Tri-n-butylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3N
CAS Number:
Molecular Weight:
185.35
Beilstein:
1698872
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
technique(s):
GC/GC: suitable
bp:
216 °C (lit.)
vapor pressure:
0.3 mmHg ( 20 °C)
2.4 mmHg ( 55 °C)

vapor density

6.38 (vs air)

Quality Level

vapor pressure

0.3 mmHg ( 20 °C)
2.4 mmHg ( 55 °C)

grade

puriss. p.a.

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

410 °F

expl. lim.

6 %

technique(s)

GC/GC: suitable

impurities

≤0.2% water

refractive index

n20/D 1.428 (lit.)
n20/D 1.429

bp

216 °C (lit.)

mp

−70 °C (lit.)

density

0.778 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

functional group

amine

SMILES string

CCCCN(CCCC)CCCC

InChI

1S/C12H27N/c1-4-7-10-13(11-8-5-2)12-9-6-3/h4-12H2,1-3H3

InChI key

IMFACGCPASFAPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tributylamine is a tertiary amine. It has been reported to prevent the corrosion of mild steel in HCl solution.

Application

Tributylamine (n-Bu3N) has been used to compose the mobile phase for the liquid chromatography tandem mass spectrometry (LC-MS/MS) analysis.
It may be used in the preparation of PdCl,N and PdOAc,N (palladium nanoparticles). It may be used as an additive during the synthesis of 3-bromothieno[3,2-c]pyridine-4-(5H)-one.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium nanoparticles obtained from palladium salts and tributylamine in molten tetrabutylammonium bromide: their use for hydrogenolysis-free hydrogenation of olefins.
Le Bras J, et al.
New. J. Chem., 28(12), 1550-1553 (2004)
Thermal Synthesis of 3-Bromothieno [3, 2-c] pyridin-4-(5H)-one: A Telescoped Procedure with Tributylamine.
Boros EE and Kaldor I.
Journal of Heterocyclic Chemistry, 52(1), 302-305 (2015)
Alexander S Misharin et al.
Analytical chemistry, 77(2), 459-470 (2005-01-15)
The analytical performance of an atmospheric pressure sampling, multiple-channel, high-throughput mass spectrometer was investigated using samples of a variety of types. The instrument, based on an array of cylindrical ion traps, was built with four independent channels and here is
A Touati et al.
Journal of protein chemistry, 11(6), 613-621 (1992-12-01)
The study of the solubility of unstructured proteins (alpha s1-, beta-, and kappa-casein) and well-structured globulin (beta-lactoglobulin) in low water binary solvent systems demonstrated the crucial importance of solvent polarity and neutralization of protein polar functions on the final outcome
I E Flesch et al.
Infection and immunity, 56(6), 1464-1469 (1988-06-01)
Bone marrow-derived murine macrophages are able to inhibit the growth of Mycobacterium bovis and of some strains of M. tuberculosis after stimulation with either recombinant gamma interferon (rIFN-gamma) or lymphokines from antigen-specific T-cell clones. To elucidate the mechanism(s) involved in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service