Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tetrahydrofuran
Synonym(s):
Butylene oxide, Oxolane, Tetramethylene oxide, Tetrahydrofuran
Empirical Formula (Hill Notation):
C
4
H
8
O
CAS Number:
109-99-9
Molecular Weight:
72.11
EC Number:
203-726-8
Products
Product Category
solvents (20)
Brand
SAFC (1)
Sigma-Aldrich (11)
Supelco (11)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
ACS reagent (5)
Anhydrous (5)
Puriss (2)
Puriss P.A. (2)
analytical standard (1)
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-20 of 24
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
34865
Tetrahydrofuran,
≥99.9%, suitable for HPLC, inhibitor-free
Expand
View Pricing
360589
Tetrahydrofuran,
≥99.0%, ACS reagent, contains 200-400 ppm BHT as inhibitor, suitable for HPLC
Expand
View Pricing
1.08101
Tetrahydrofuran,
≥99.9% (GC), suitable for HPLC, LiChrosolv
®
Expand
View Pricing
5.89570
Tetrahydrofuran,
≥99.9%, anhydrous, contains 250 ppm BHT as inhibitor
Expand
View Pricing
1.09731
Tetrahydrofuran,
≥99.8% (GC), ACS reagent, reag. Ph. Eur., EMSURE
®
Expand
View Pricing
178810
Tetrahydrofuran,
≥99.0%, contains 200-400 ppm BHT as inhibitor,
ReagentPlus
®
Expand
View Pricing
87368
Tetrahydrofuran,
≥99.9%, ACS reagent, contains ~0.025% 2, 6-di-tert-butyl-4-methylphenol as stabilizer, 250 ppm BHT as inhibitor, suitable for solid phase extraction (SPE), reag. Ph. Eur.
Expand
View Pricing
5.89568
Tetrahydrofuran,
≥99.9%, anhydrous, inhibitor-free
Expand
View Pricing
676764
Tetrahydrofuran,
≥99.0%, ACS reagent, contains 200-400 ppm BHT as inhibitor, suitable for HPLC
Expand
View Pricing
401757
Tetrahydrofuran,
≥99.9%, anhydrous, inhibitor-free
Expand
View Pricing
186562
Tetrahydrofuran,
≥99.9%, anhydrous, contains 250 ppm BHT as inhibitor
Expand
View Pricing
1.08114
Tetrahydrofuran,
≥99% (GC), EMPLURA
®
Expand
View Pricing
1.07025
Tetrahydrofuran,
≥99.5% (GC), EMPARTA
®
ACS reagent
Expand
View Pricing
270385
Tetrahydrofuran,
≥99.9%, HPLC grade, suitable for HPLC, inhibitor-free
Expand
View Pricing
TX0280
Tetrahydrofuran,
≥99.5% (GC), Meets ACS Specifications, General Reagents grade, contains 0.0140-0.0310% BHT as stabilizer
Expand
View Pricing
1.08110
Tetrahydrofuran,
≥99.9% (GC), suitable for UV/Vis spectroscopy, Uvasol
®
Expand
View Pricing
1.08107
Tetrahydrofuran,
≥99.9% (GC), suitable for DNA amplification and peptide synthesis (max. 0.005% H₂O)
Expand
View Pricing
1.37136
Tetrahydrofuran,
EMPROVE
®
ESSENTIAL
Expand
View Pricing
87369
Tetrahydrofuran,
≥99.5%, ≥99.5% (GC), for ion-selective electrodes, contains 250 ppm BHT as inhibitor, Selectophore
™
Expand
View Pricing
78445
Tetrahydrofuran,
analytical standard
Expand
View Pricing
Page 1 of 2
Page 1 of 2