Skip to Content
Merck
All Photos(1)

Documents

77460

Sigma-Aldrich

2-Chloroacetophenone

purum, ≥98.0% (GC)

Synonym(s):

ω-Chloroacetophenone, Phenacyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH2Cl
CAS Number:
Molecular Weight:
154.59
Beilstein:
507950
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (GC)

form

crystals

bp

244-245 °C (lit.)

mp

53-56 °C
54-56 °C (lit.)

density

1.324 g/mL at 25 °C (lit.)

SMILES string

ClCC(=O)c1ccccc1

InChI

1S/C8H7ClO/c9-6-8(10)7-4-2-1-3-5-7/h1-5H,6H2

InChI key

IMACFCSSMIZSPP-UHFFFAOYSA-N

Gene Information

human ... PTPN6(5777)

Looking for similar products? Visit Product Comparison Guide

General description

2-Chloroacetophenone is an organic building block used in the synthesis of heterocyclic compounds. It has low solubility in water but can be soluble in alcohols, ethers and other organic solvents.

Application

2-Chloroacetophenone can be used in the synthesis of phenyl acetic acid by Photo-Favorskii rearrangement. It is also used in the preparation of 1-(2-chlorophenyl)ethanol via asymmetric hydrogenation reaction.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of microgel-supported chiral catalysts and their application in the asymmetric hydrogenation of aromatic ketones
Deng J, et al.
Reactive and Functional Polymers, 72, 378-382 (2012)
Principles of coaxial illumination for photochemical reactions: Part 2. Model validation
Meir, et al.
Journal of advanced materials and processing, 2 (2020)
Ionic liquid mediated and promoted one-pot green synthesis of new isoxazolyl dihydro-1 H-indol-4 (5 H)-one derivatives at ambient temperature
Modugu, et al.
Cogent Chemistry , 3, 1318693-1318693 (2017)
Experimental study of the adsorption of 2-Chloroacetophenone at the air-environmental water interface
Hao, et al.
Frontiers in environmental science., 10, 2254-2254 (2022)
Bert Brône et al.
Toxicology and applied pharmacology, 231(2), 150-156 (2008-05-27)
The TRPA1 channel is activated by a number of pungent chemicals, such as allylisothiocyanate, present in mustard oil and thiosulfinates present in garlic. Most of the known activating compounds contain reactive, electrophilic chemical groups, reacting with cysteine residues in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service