Skip to Content
Merck
All Photos(1)

Documents

C19686

Sigma-Aldrich

2-Chloroacetophenone

98%

Synonym(s):

ω-Chloroacetophenone, Phenacyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH2Cl
CAS Number:
Molecular Weight:
154.59
Beilstein:
507950
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

bp

244-245 °C (lit.)

mp

54-56 °C (lit.)

density

1.324 g/mL at 25 °C (lit.)

SMILES string

ClCC(=O)c1ccccc1

InChI

1S/C8H7ClO/c9-6-8(10)7-4-2-1-3-5-7/h1-5H,6H2

InChI key

IMACFCSSMIZSPP-UHFFFAOYSA-N

Gene Information

human ... PTPN6(5777)

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Efficient whole-cell biotransformation in a biphasic ionic liquid/water system.
Holger Pfruender et al.
Angewandte Chemie (International ed. in English), 43(34), 4529-4531 (2004-09-02)
Bert Brône et al.
Toxicology and applied pharmacology, 231(2), 150-156 (2008-05-27)
The TRPA1 channel is activated by a number of pungent chemicals, such as allylisothiocyanate, present in mustard oil and thiosulfinates present in garlic. Most of the known activating compounds contain reactive, electrophilic chemical groups, reacting with cysteine residues in the
Lenilson C Rocha et al.
Biotechnology letters, 31(10), 1559-1563 (2009-06-06)
The asymmetric reduction of 2-chloro-1-phenylethanone (1) by seven strains of marine fungi was evaluated and afforded (S)-(-)-2-chloro-1-phenylethanol with, in the best case, an enantiomeric excess of 50% and an isolated yield of 60%. The ability of marine fungi to catalyse
Anil K Nigam et al.
Journal of hazardous materials, 184(1-3), 506-514 (2010-09-17)
Pyrolysis-GC/MS system with on-line micro-furnace was used to make rapid evaluation of ω-chloroacetophenone (CN) decomposition under inert thermal atmospheres. The volatile products evolved during pyrolysis were analyzed by thermal gravimetric analysis (TGA) and Py-GC/MS to obtain specific thermogram and pyrogram.
Wladimir Solodenko et al.
Molecular diversity, 9(4), 333-339 (2005-11-29)
The preparation of a new palladium(II) complex with a 2-pyridinealdoxime ligand and its use as a Pd(0) precatalyst in the cross-coupling Suzuki-Miyaura reaction is described. Several concepts for the immobilization of this catalytic system are presented and compared in order

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service