Skip to Content
Merck
All Photos(1)

Key Documents

71325

Supelco

Sodium bromate

puriss. p.a., ≥99.5% (RT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBrO3
CAS Number:
Molecular Weight:
150.89
EC Number:
MDL number:
UNSPSC Code:
12000000
eCl@ss:
38050507
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.5% (RT)
form:
crystals

grade

puriss. p.a.

Quality Level

Assay

≥99.5% (RT)

form

crystals

impurities

≤50 mg/kg total nitrogen (N)

pH

5.0-9.0 (25 °C, 50 mg/mL in H2O)

density

3.339 g/mL at 25 °C (lit.)

anion traces

bromide (Br-): ≤500 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤100 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Na+].[O-]Br(=O)=O

InChI

1S/BrHO3.Na/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

XUXNAKZDHHEHPC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium bromate is an organic compound which is sodium salt of bromic acid. This alkali metal bromate is mostly used as oxidizing agents.

Application

Sodium bromate was used in gel media preparation to understand its crystal growth. It was also used to prepare aqueous anion solution.

Other Notes

Cooxidant for a catalytic cerium(IV) oxidation procedure

Pictograms

Flame over circleSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Ox. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Water quality factors affecting bromate reduction in biologically active carbon filters.
Kirisits, Mary Jo, et al.
Water Research, 35.4, 891-900 (2001)
Pradyot Patnaik
A Comprehensive Guide to the Hazardous Properties of Chemical Substances, 709-709 (2007)
T.-L. Ho
Synthetic Communications, 9, 237-237 (1979)
Habit changes of sodium bromate crystals grown from gel media.
Petrova, Rositza I., and Jennifer A.
Crystal Growth & Design, 2.6, 573-578 (2002)
S. Kanemoto et al.
Bulletin of the Chemical Society of Japan, 59, 105-105 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service