Skip to Content
Merck
All Photos(4)

Documents

L6402

Sigma-Aldrich

L-(+)-Lactic acid

BioXtra, ≥98% (titration)

Synonym(s):

(S)-2-Hydroxypropionic acid, Sarcolactic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
Beilstein:
1720251
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Assay

≥98% (titration)

impurities

≤0.005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

antibiotic activity spectrum

Gram-negative bacteria

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

C[C@H](O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m0/s1

InChI key

JVTAAEKCZFNVCJ-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

L-(+)-Lactic acid is used as a substrate for lactic acid dehydrogenase and lactate oxidase.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia Selleri et al.
Oncotarget, 7(21), 30193-30210 (2016-04-14)
Human mesenchymal stromal cells (MSC) have been shown to dampen immune response and promote tissue repair, but the underlying mechanisms are still under investigation. Herein, we demonstrate that umbilical cord-derived MSC (UC-MSC) alter the phenotype and function of monocyte-derived dendritic
Melissa M Keenan et al.
PLoS genetics, 11(10), e1005599-e1005599 (2015-10-10)
In order to propagate a solid tumor, cancer cells must adapt to and survive under various tumor microenvironment (TME) stresses, such as hypoxia or lactic acidosis. To systematically identify genes that modulate cancer cell survival under stresses, we performed genome-wide
Koen Poesen et al.
Clinical chemistry and laboratory medicine, 51(7), 1417-1427 (2013-03-16)
With the use of a traditional blood gas analyzer (BGA) (ABL800 Radiometer) for blood glucose monitoring, tight glucose control (TGC) reduced in-hospital mortality and morbidity of surgical intensive care unit (ICU) adult and pediatric patients. Such BGAs are now superseded
David M Garcia et al.
eLife, 5 (2016-12-03)
Robust preference for fermentative glucose metabolism has motivated domestication of the budding yeast Saccharomyces cerevisiae. This program can be circumvented by a protein-based genetic element, the [GAR+] prion, permitting simultaneous metabolism of glucose and other carbon sources. Diverse bacteria can
Shanjie Wang et al.
Journal of cellular and molecular medicine, 22(10), 5132-5144 (2018-08-01)
Mitophagy eliminates dysfunctional mitochondria and thus plays a cardinal role in diabetic cardiomyopathy (DCM). We observed the favourable effects of melatonin on cardiomyocyte mitophagy in mice with DCM and elucidated their underlying mechanisms. Electron microscopy and flow cytometric analysis revealed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service