Skip to Content
Merck
All Photos(1)

Documents

69745

Supelco

δ-Tocotrienol

analytical standard

Synonym(s):

(R)-δ-Tocotrienol, 8-Methyltocotrienol, [R-(E,E)]-3,4-Dihydro-2,8-dimethyl-2-(4,8,12-trimethyl-3,7,11-tridecatrienyl)-2H-1-benzopyran-6-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H40O2
CAS Number:
Molecular Weight:
396.61
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

biological source

Elaeis guineensis

Quality Level

grade

analytical standard

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

colorless to amber

application(s)

pharmaceutical (small molecule)
vitamins, nutraceuticals, and natural products

format

neat

storage temp.

−20°C

SMILES string

C\C(C)=C\CC\C(C)=C\CC\C(C)=C\CC[C@]1(C)CCc2cc(O)cc(C)c2O1

InChI

1S/C27H40O2/c1-20(2)10-7-11-21(3)12-8-13-22(4)14-9-16-27(6)17-15-24-19-25(28)18-23(5)26(24)29-27/h10,12,14,18-19,28H,7-9,11,13,15-17H2,1-6H3/b21-12+,22-14+/t27-/m1/s1

InChI key

ODADKLYLWWCHNB-LDYBVBFYSA-N

Related Categories

General description

δ-Tocotrienol can be found in coconut oil, palm oil, and cereal grains such as barley, oats, wheat and rye.
δ-Tocotrienol is known as one of the homologs of Vitamin E.

Application

δ-Tocotrienol may be used as an analytical reference standard for the determination of δ-tocotrienol in:
  • Food samples of vegetable origin by high performance liquid chromatography-fluorescene detection (HPLC-FLD).
  • Chicken meat samples by LC-FLD.
  • Cereals by normal phase HPLC-FLD.
  • Rice bran samples by one-step equilibrium extraction method coupled to reversed-phase HPLC combined with dual wavelength ultraviolet-visible (UV-Vis) detector.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A rapid procedure for analysing rice bran tocopherol, tocotrienol and gamma-oryzanol contents.
Chen MH and Bergman CJ
J. Food Compos. Anal., 18(2-3), 139-151 (2005)
Determination of carotene, tocopherols and tocotrienols in residue oil from palm pressed fiber using pressurized liquid extraction-normal phase liquid chromatography
Sanagi MM, et al.
Analytica Chimica Acta, 538(1-2), 71-76 (2005)
Merriline M Satyamitra et al.
Radiation research, 175(6), 736-745 (2011-03-26)
δ-Tocotrienol (DT3), a vitamin E isoform, is associated with strong antioxidant and immunomodulatory properties. We confirmed the potent antioxidant activity in membrane systems and showed that DT3 is an effective radiation protector and mitigator. DT3 (4 μM, P < 0.001)
Method for simultaneous analysis of eight vitamin E isomers in various foods by high performance liquid chromatography and fluorescence detection.
Pinheiro-Sant?Ana HM, et al.
Journal of Chromatography A, 1218(47), 8496-8502 (2011)
Asaf A Qureshi et al.
Lipids in health and disease, 10, 239-239 (2011-12-22)
Changes in immune function believed to contribute to a variety of age-related diseases have been associated with increased production of nitric oxide (NO). We have recently reported that proteasome inhibitors (dexamethasone, mevinolin, quercetin, δ-tocotrienol, and riboflavin) can inhibit lipopolysaccharide (LPS)-induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service