Skip to Content
Merck
All Photos(1)

Documents

05644

Supelco

β-Tocotrienol

analytical standard

Synonym(s):

(R)-β-Tocotrienol, [R-(E,E)]-3,4-Dihydro-2,5,8-trimethyl-2-(4,8,12-trimethyl-3,7,11-tridecatrienyl)-2H-1-benzopyran-6-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H42O2
CAS Number:
Molecular Weight:
410.63
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

biological source

Elaeis guineensis

Quality Level

grade

analytical standard

Assay

≥95.0% (HPLC)

form

liquid
viscous liquid

shelf life

limited shelf life, expiry date on the label

color

colorless to amber

application(s)

vitamins, nutraceuticals, and natural products

format

neat

storage temp.

−20°C

SMILES string

C\C(C)=C\CC\C(C)=C\CC\C(C)=C\CC[C@]1(C)CCc2c(C)c(O)cc(C)c2O1

InChI

1S/C28H42O2/c1-20(2)11-8-12-21(3)13-9-14-22(4)15-10-17-28(7)18-16-25-24(6)26(29)19-23(5)27(25)30-28/h11,13,15,19,29H,8-10,12,14,16-18H2,1-7H3/b21-13+,22-15+/t28-/m1/s1

InChI key

FGYKUFVNYVMTAM-WAZJVIJMSA-N

General description

β-Tocotrienol is a naturally occurring forms of vitamin E commonly found in cereals such as whole wheat flour.

Application

β-Tocotrienol may be used as an analytical reference standard for the quantification of the analyte in chicken meat samples using liquid chromatography technique.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Yu et al.
Nutrition and cancer, 33(1), 26-32 (1999-05-05)
The apoptosis-inducing properties of RRR-alpha-, beta-, gamma-, and delta-tocopherols, alpha-, gamma-, and delta-tocotrienols, RRR-alpha-tocopheryl acetate (vitamin E acetate), and RRR-alpha-tocopheryl succinate (vitamin E succinate) were investigated in estrogen-responsive MCF7 and estrogen-nonresponsive MDA-MB-435 human breast cancer cell lines in culture. Apoptosis
V Piironen et al.
International journal for vitamin and nutrition research. Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Journal international de vitaminologie et de nutrition, 54(1), 35-40 (1984-01-01)
A HPLC Method is described for the determination of tocopherols and tocotrienols in human diets and plasma. After a room-temperature saponification diet samples were extracted with n-hexane. A direct hexane extraction was used for plasma samples. Using a normal-phase column
E J Konings et al.
Journal of AOAC International, 79(4), 902-906 (1996-07-01)
A liquid chromatographic (LC) method was developed for determination of tocopherols and tocotrienols in foods. Tocopherols and tocotrienols were released after saponification of test portions for 40 min at 80 degrees C, followed by extraction with hexane-ethyl acetate. After evaporation
Grain-based Foods: Processing, Properties, and Heath Attributes (2018)
P F Surai et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 121(4), 393-396 (1999-02-11)
The aim of this study was to investigate the possibility of biodiscrimination between different forms of vitamin E during the development of the chick embryo. The vitamin E present in the initial yolk consisted of alpha-tocopherol (90%), (beta + gamma)-tocopherol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service