Skip to Content
Merck
All Photos(3)

Key Documents

48710

Sigma-Aldrich

Propyl gallate

≥98.0% (HPLC), solid, antioxidant

Synonym(s):

3,4,5-Trihydroxybenzoic acid propyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3,4,5-(HO)3C6H2CO2CH2CH2CH3
CAS Number:
Molecular Weight:
212.20
Beilstein:
1877976
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
39024506
PubChem Substance ID:
NACRES:
NA.77

Product Name

Propyl gallate, antioxidant, ≥98.0% (HPLC)

Quality Level

Assay

≥98.0% (HPLC)

form

solid

mp

146-149 °C (lit.)
146-149 °C

SMILES string

CCCOC(=O)c1cc(O)c(O)c(O)c1

InChI

1S/C10H12O5/c1-2-3-15-10(14)6-4-7(11)9(13)8(12)5-6/h4-5,11-13H,2-3H2,1H3

InChI key

ZTHYODDOHIVTJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Reconstitution

Prepare a 0.1M solution in glycerol:PBS (9:1)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

368.6 °F - closed cup

Flash Point(C)

187 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matloob Ahmad et al.
European journal of medicinal chemistry, 45(2), 698-704 (2009-12-08)
A series of potential anti-oxidant and anti-bacterial N'-arylmethylidene-2-(3,4-dimethyl-5,5-dioxidopyrazolo[4,3-c][1,2]benzothiazin-2(4H)-yl)acetohydrazides was synthesized in a facile way starting from commercially available saccharine. 3,4-Dimethyl-2,4-dihydropyrazolo[4,3-c][1,2]benzothiazine 5,5-dioxide was obtained by ring expansion of 2-(2-oxopropyl)-1,2-benzisothiazol-3(2H)-one 1,1-dioxide followed by N-methylation and cyclization with hydrazine using ultrasonic irradiation. N-alkylation
Paola Colindres et al.
Journal of the science of food and agriculture, 91(5), 963-968 (2011-01-22)
The effect of selected antioxidants (grape seed extract (GS), oleoresin rosemary (OR), water-soluble oregano extract (WO), propyl gallate (PG), butylated hydroxyanisole (BHA)), butylated hydroxytoluene (BHT)) on sensory, color and oxidative stability of cooked, frozen, reheated ground beef patties was evaluated.
Brianne A Wilton et al.
Virology, 374(1), 82-99 (2008-01-29)
Cellular proteins containing BTB and kelch domains have been shown to function as adapters for the recruitment of substrates to cullin-3-based ubiquitin ligases. Poxviruses are the only family of viruses known to encode multiple BTB/kelch proteins, suggesting that poxviruses may
Sonia Losada-Barreiro et al.
Journal of colloid and interface science, 389(1), 1-9 (2012-09-04)
We evaluated the effects of the hydrophile-lipophile balance (HLB) and emulsifier concentration on the distribution of the antioxidants gallic acid (GA), propyl gallate (PG), and α-tocopherol (TOC) between the aqueous, interfacial, and oil regions of food-grade emulsions composed of stripped
Qi Chen et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(1), 118-130 (2009-01-09)
Axonal dysfunction is the major phenotypic change in many neurodegenerative diseases, but the processes underlying this impairment are not clear. Modifier of cell adhesion (MOCA) is a presenilin binding protein that functions as a guanine nucleotide exchange factor for Rac1.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service