Skip to Content
Merck
All Photos(1)

Documents

02370

Sigma-Aldrich

Propyl gallate

for microscopy, ≥98.0% (HPLC)

Synonym(s):

3,4,5-Trihydroxybenzoic acid propyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3,4,5-(HO)3C6H2CO2CH2CH2CH3
CAS Number:
Molecular Weight:
212.20
Beilstein:
1877976
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39024506
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy

Assay

≥98.0% (HPLC)

form

powder

pH range

5.7-6.3

mp

146-149 °C (lit.)

density

1.21 g/cm3

application(s)

diagnostic assay manufacturing
hematology
histology

SMILES string

CCCOC(=O)c1cc(O)c(O)c(O)c1

InChI

1S/C10H12O5/c1-2-3-15-10(14)6-4-7(11)9(13)8(12)5-6/h4-5,11-13H,2-3H2,1H3

InChI key

ZTHYODDOHIVTJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Propyl gallate is used as an anti-fade reagent in fluorescence microscopy to reduce photobleaching of fluorescent probes.

Reconstitution

Prepare a 0.1M solution in glycerol:PBS (9:1)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

368.6 °F - closed cup

Flash Point(C)

187 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Blassan Plackal Adimuriyil George et al.
Scientific reports, 8(1), 14368-14368 (2018-09-27)
Metallic nanoparticles (NPs) especially silver (Ag) NPs have shown immense potential in medical applications due to their distinctive physio-chemical and biological properties. This article reports the conjugation of Ag NPs with Rubus fairholmianus extract. The modification of Ag NPs was
Lynn M Boyden et al.
The Journal of investigative dermatology, 135(6), 1540-1547 (2014-11-15)
Genetic investigation of inherited skin disorders has informed the understanding of skin self-renewal, differentiation, and barrier function. Erythrokeratodermia variabilis et progressiva (EKVP) is a rare, inherited skin disease that is characterized by transient figurate patches of erythema, localized or generalized
Pablo Rougerie et al.
Scientific reports, 10(1), 14784-14784 (2020-09-10)
How biophysical cues can control tissue morphogenesis is a central question in biology and for the development of efficient tissue engineering strategies. Recent data suggest that specific topographies such as grooves and ridges can trigger anisotropic tissue growth. However, the
Yasunobu Kawano et al.
Resuscitation, 83(2), 249-252 (2011-08-02)
The present study was conducted to assess the effects of intraperitoneal administration of n-propyl gallate (PG) on hippocampal neuronal survival after forebrain ischemia. Forty male Sprague-Dawley rats were randomly assigned to one of 6 groups. Animals in the PG-I-10, PG-I-8
Brianne A Wilton et al.
Virology, 374(1), 82-99 (2008-01-29)
Cellular proteins containing BTB and kelch domains have been shown to function as adapters for the recruitment of substrates to cullin-3-based ubiquitin ligases. Poxviruses are the only family of viruses known to encode multiple BTB/kelch proteins, suggesting that poxviruses may

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service