Skip to Content
Merck
All Photos(1)

Key Documents

35952

Supelco

Phenol

PESTANAL®, analytical standard

Synonym(s):

Hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
Beilstein:
969616
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.24 (vs air)

vapor pressure

0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

product line

PESTANAL®

autoignition temp.

1319 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

8.6 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

182 °C (lit.)

mp

40-42 °C (lit.)

density

1.071 g/mL at 25 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Britta T Eklund et al.
Marine pollution bulletin, 46(2), 171-181 (2003-02-15)
Toxic effects on macroalgae have been compiled. Eighty-two articles have been found in literature during 1959-2000. A total of 120 substances were investigated using 65 different macroalgae species. About one-third of the tested compounds were organic substances (33%), another third
Serdar Durdagi et al.
Bioorganic & medicinal chemistry, 19(4), 1381-1389 (2011-02-02)
Carbonic anhydrases (CAs, EC 4.2.1.1) are inhibited by sulfonamides, inorganic anions, phenols, coumarins (acting as prodrugs) and polyamines. A novel class of CA inhibitors (CAIs), interacting with the CA isozymes I, II (cytosolic) and IX, XII (transmembrane, tumor-associated) in a
Yukari Yamane-Sando et al.
MicrobiologyOpen, 3(2), 196-212 (2014-02-11)
Sphingolipids are a family of eukaryotic lipids biosynthesized from sphingoid long-chain bases (LCBs). Sphingolipids are an essential class of lipids, as their depletion results in cell death. However, acute LCB supplementation is also toxic; thus, proper cellular LCB levels should
Fabrizio Carta et al.
Journal of medicinal chemistry, 53(15), 5511-5522 (2010-07-02)
Carbonic anhydrases (CAs, EC 4.2.1.1) are inhibited by sulfonamides, phenols, and coumarins. Polyamines such as spermine, spermidine, and many synthetic congeners are described to constitute a novel class of CA inhibitors (CAIs), interacting with the different CA isozymes with efficiency
Patrick W Causey et al.
Journal of medicinal chemistry, 51(9), 2833-2844 (2008-04-17)
A series of mono and diaryl rhenium(I)-carborane derivatives were prepared using microwave heating and screened for their affinity for two isoforms of the estrogen receptor (ER). The rhenacarborane derivative [(RR'C 2B9H9)Re(CO)3](-) (R = p-PhOH, R' = H), which was generated

Protocols

Analytical standard separation of various phenols for research and industrial applications.

Analytical standard separation of various phenols for research and industrial applications.

Analytical standard separation of various phenols for research and industrial applications.

Analytical standard separation of various phenols for research and industrial applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service