Skip to Content
Merck
All Photos(1)

Key Documents

45835

Supelco

Cypermethrin solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19Cl2NO3
CAS Number:
Molecular Weight:
416.30
Beilstein:
8444118
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

mixture of isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

SMILES string

CC1(C)C(\C=C(\Cl)Cl)C1C(=O)OC(C#N)c2cccc(Oc3ccccc3)c2

InChI

1S/C22H19Cl2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3

InChI key

KAATUXNTWXVJKI-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Waqas Wakil et al.
PloS one, 13(7), e0192628-e0192628 (2018-07-20)
The red palm weevil Rhynchophorus ferrugineus (Olivier) is an important pest of date palms in many regions of the world. This paper reports the first survey of insecticide resistance in field populations of R. ferrugineus in Pakistan which were collected
Franco Andrés Pascucci et al.
Biochimica et biophysica acta. Molecular cell research, 1868(3), 118918-118918 (2020-12-07)
Normal-to-tumor cell transition is accompanied by changes in gene expression and signal transduction that turns the balance toward cancer-cell phenotype, eluding by different mechanisms, the response of tumor-suppressor genes. Here, we observed that MageC2, a MAGE-I protein able to regulate
Muthuviveganandavel Veerappan et al.
International journal of experimental pathology, 93(5), 361-369 (2012-09-15)
Alpha-cypermethrin and carbendazim are synthetic; α-cypermethrin belongs to a class of synthetic pyrethroids and carbendazim belongs to the class of carbamate fungicides. The current study was carried out to evaluate the low-dose exposure of individual and mixed forms of cypermethrin
Richard M Oxborough et al.
PloS one, 8(2), e55781-e55781 (2013-02-15)
Pyrethroid resistant Anopheles gambiae malaria vectors are widespread throughout sub-Saharan Africa and continued efficacy of pyrethroid ITNs is under threat. Chlorfenapyr is a promising pyrrole insecticide with a unique mechanism of action conferring no cross-resistance to existing public health insecticides.
Yan Fang Li et al.
Biomedical and environmental sciences : BES, 26(3), 201-208 (2013-02-22)
To evaluate effects of cypermethrin on the testis histology and testosterone, LH and FSH in adult male Sprague-Dawley rats. The intact adult male rats were randomly divided into five groups and were treated with cypermethrin at doses of 0, 7.5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service