Skip to Content
Merck
All Photos(1)

Key Documents

45626

Supelco

Piperonylbutoxide

PESTANAL®, analytical standard

Synonym(s):

2-(2-Butoxyethoxy)ethyl (6-propylpiperonyl) ether, 4,5-Methylenedioxy-2-propylbenzyldiethyleneglycol butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O5
CAS Number:
Molecular Weight:
338.44
Beilstein:
288063
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.498 (lit.)

bp

155 °C/0.3 mmHg (lit.)

density

1.059 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCCCOCCOCCOCc1cc2OCOc2cc1CCC

InChI

1S/C19H30O5/c1-3-5-7-20-8-9-21-10-11-22-14-17-13-19-18(23-15-24-19)12-16(17)6-4-2/h12-13H,3-11,14-15H2,1-2H3

InChI key

FIPWRIJSWJWJAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

339.8 °F - closed cup

Flash Point(C)

171 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Petek Piner et al.
Environmental toxicology and pharmacology, 33(3), 414-420 (2012-03-06)
The aim of this study was to investigate the toxic effects of pyrethroid pesticide lambda-cyhalothrin in the presence of piperonyl butoxide as a modulator in the liver of juvenile Oreochromis niloticus. LC(50) (96h) value of lambda-cyhalothrin was determined as 2.901μg/L
Olivier J T Briët et al.
Malaria journal, 12, 77-77 (2013-02-28)
The effectiveness of insecticide-treated nets in preventing malaria is threatened by developing resistance against pyrethroids. Little is known about how strongly this affects the effectiveness of vector control programmes. Data from experimental hut studies on the effects of long-lasting, insecticidal
Rodolphe Poupardin et al.
PLoS neglected tropical diseases, 8(3), e2743-e2743 (2014-03-22)
Thailand is currently experiencing one of its worst dengue outbreaks in decades. As in most countries where this disease is endemic, dengue control in Thailand is largely reliant on the use of insecticides targeting both immature and adult stages of
N Karatolos et al.
Insect molecular biology, 21(3), 327-334 (2012-03-31)
Spiromesifen is a novel insecticide and is classed as a tetronic acid derivative. It targets the insects' acetyl-coenzyme A carboxylase (ACCase) enzyme, causing a reduction in lipid biosynthesis. At the time of this publication, there are no reports of resistance
Despina Philippou et al.
Pest management science, 69(4), 499-506 (2012-09-13)
It has been reported previously that piperonyl butoxide (PBO) can inhibit both P450 and esterase activity. Although the method by which PBO combines with cytochrome P450 has been identified, the way in which it acts as an esterase inhibitor has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service