Skip to Content
Merck
All Photos(1)

Key Documents

45640

Supelco

Propazine

PESTANAL®, analytical standard

Synonym(s):

2,4-Bis(isopropylamino)-6-chloro-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16ClN5
CAS Number:
Molecular Weight:
229.71
Beilstein:
747081
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CC(C)Nc1nc(Cl)nc(NC(C)C)n1

InChI

1S/C9H16ClN5/c1-5(2)11-8-13-7(10)14-9(15-8)12-6(3)4/h5-6H,1-4H3,(H2,11,12,13,14,15)

InChI key

WJNRPILHGGKWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propazine is a triazine herbicide, used in controlling broad-leaved weeds and annual grasses in sweet sorghum.

Application

Propazine may be used as a reference standard for the determination of propazine in water samples using enzyme-linked Immunosorbent assay (ELISA) and gas chromatography-mass spectrometry detection (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pesticide Profiles: Toxicity, Environmental Impact, and Fate (1997)
Enzyme-linked immunosorbent assay compared with gas chromatography/mass spectrometry for the determination of triazine herbicides in water.
Thurman ME, et al.
Analytical Chemistry, 62(18), 2043-2048 (1990)
Pesticide Profiles: Toxicity, Environmental Impact, and Fate (1997)
H Jiang et al.
Environmental science & technology, 40(11), 3609-3616 (2006-06-22)
Atrazine, simazine, and propazine and their major chlorinated degradates (deethylatrazine, deisopropylatrazine, and didealkylatrazine) are considered as a group to be endocrine-disrupting chemicals by the U.S. Environmental Protection Agency. On this basis, regulatory action levels are currently under consideration for the
Luciana B O Dos Santos et al.
Journal of separation science, 32(4), 494-500 (2009-02-13)
This paper describes the development of a sequential injection chromatography (SIC) procedure for separation and quantification of the herbicides simazine, atrazine, and propazine exploring the low backpressure of a 2.5 cm long monolithic C(18) column. The separation of the three

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service