Skip to Content
Merck
All Photos(1)

Documents

45533

Supelco

3-Indoleacetic acid

PESTANAL®, analytical standard

Synonym(s):

Heteroauxin, IAA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
Beilstein:
143358
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

98%

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

165-169 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)Cc1c[nH]c2ccccc12

InChI

1S/C10H9NO2/c12-10(13)5-7-6-11-9-4-2-1-3-8(7)9/h1-4,6,11H,5H2,(H,12,13)

InChI key

SEOVTRFCIGRIMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Indoleacetic acid is a highly effective, growth promotor in lower plant life, formed by a variety of fungi, including yeast and has been isolated from Aspergillus niger and Rhizopus sp. It is commonly employed in horticulture and industry.

Application

3-Indoleacetic acid may be used as an analytical reference standard for the quantification of the analyte in environmental samples using gas chromatography coupled to mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiresidue analysis of pollutants as their trimethylsilyl derivatives, by gas chromatography-mass spectrometry
Sebok A, et al.
Journal of Chromatography A, 1216(12), 2288-2301 (2009)
Humic Matter in Soil and the Environment: Principles and Controversies (2003)
Amir H Ahkami et al.
Planta, 238(3), 499-517 (2013-06-15)
To determine the contribution of polar auxin transport (PAT) to auxin accumulation and to adventitious root (AR) formation in the stem base of Petunia hybrida shoot tip cuttings, the level of indole-3-acetic acid (IAA) was monitored in non-treated cuttings and
Ana Zúñiga et al.
Molecular plant-microbe interactions : MPMI, 26(5), 546-553 (2013-01-11)
Although not fully understood, molecular communication in the rhizosphere plays an important role regulating traits involved in plant-bacteria association. Burkholderia phytofirmans PsJN is a well-known plant-growth-promoting bacterium, which establishes rhizospheric and endophytic colonization in different plants. A competent colonization is
Xinhua Dai et al.
The Journal of biological chemistry, 288(3), 1448-1457 (2012-11-29)
Auxin regulates every aspect of plant growth and development. Previous genetic studies demonstrated that YUCCA (YUC) flavin-containing monooxygenases (FMOs) catalyze a rate-limiting step in auxin biosynthesis and that YUCs are essential for many developmental processes. We proposed that YUCs convert

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service