Skip to Content
Merck
All Photos(1)

Key Documents

35745

Supelco

1-Naphthaleneacetic acid

PESTANAL®, analytical standard

Synonym(s):

α-Naphthaleneacetic acid, 1-Naphthylacetic acid, NAA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10O2
CAS Number:
Molecular Weight:
186.21
Beilstein:
1308415
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

95-100% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)Cc1cccc2ccccc12

InChI

1S/C12H10O2/c13-12(14)8-10-6-3-5-9-4-1-2-7-11(9)10/h1-7H,8H2,(H,13,14)

InChI key

PRPINYUDVPFIRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Naphthaleneacetic acid is widely employed in agriculture as a synthetic plant hormone in the auxin family or a plant growth regulator, which can find applications in tissue culture.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flow-through optosensing of 1-naphthaleneacetic acid in water and apples by heavy atom induced-room temperature phosphorescence measurements
Fernandez-Arguelles TM, et al.
Talanta, 66(3), 696-702 (2005)
Auxins Regulations of Branched Spike Development and Expression of TFL, a LEAFY-Like Gene in Branched Spike Wheat (Triticum aestivum)
Yue W, et al.
Journal of Agricultural Science, 9(2), 27-27 (2017)
Da Xiao et al.
Biochemical pharmacology, 84(2), 232-239 (2012-04-20)
Carboxylesterases (CES) constitute a class of hydrolytic enzymes that play critical roles in drug metabolism and lipid mobilization. Previous studies with a large number of human liver samples have suggested that the inducibility of carboxylesterases is inversely related with age.
Peter Marhavý et al.
The EMBO journal, 32(1), 149-158 (2012-11-28)
Lateral root (LR) formation is initiated when pericycle cells accumulate auxin, thereby acquiring founder cell (FC) status and triggering asymmetric cell divisions, giving rise to a new primordium. How this auxin maximum in pericycle cells builds up and remains focused
Daeshik Cho et al.
Plant physiology, 160(3), 1293-1302 (2012-08-31)
CATION EXCHANGERs CAX1 and CAX3 are vacuolar ion transporters involved in ion homeostasis in plants. Widely expressed in the plant, they mediate calcium transport from the cytosol to the vacuole lumen using the proton gradient across the tonoplast. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service