Skip to Content
Merck
All Photos(1)

Documents

45522

Supelco

Hexachlorobenzene

PESTANAL®, analytical standard

Synonym(s):

HCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Cl6
CAS Number:
Molecular Weight:
284.78
Beilstein:
1912585
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

323-326 °C (lit.)

mp

227-229 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl

InChI

1S/C6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9

InChI key

CKAPSXZOOQJIBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - STOT RE 1 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manas K Panda et al.
Nature chemistry, 7(1), 65-72 (2014-12-18)
The exceptional mechanical flexibility observed with certain organic crystals defies the common perception of single crystals as brittle objects. Here, we describe the morphostructural consequences of plastic deformation in crystals of hexachlorobenzene that can be bent mechanically at multiple locations
C C Michielsen et al.
Environmental health perspectives, 107 Suppl 5, 783-792 (1999-09-30)
Hexachlorobenzene (HCB) is a persistent environmental pollutant. The toxicity of HCB has been extensively studied after an accidental human poisoning in Turkey and more recently it has been shown that HCB has immunotoxic properties in laboratory animals and probably also
A P van Birgelen
Environmental health perspectives, 106(11), 683-688 (1998-11-03)
A dioxinlike compound is a compound that binds to the aryl hydrocarbon (Ah) receptor, results in dioxinlike effects, and bioaccumulates. These are the three factors for including dioxinlike chemicals in the toxic equivalency factor (TEF) concept. Risk assessment of dioxinlike
Guang Wang et al.
Environment international, 36(1), 122-130 (2009-10-13)
This article summarizes the published scientific data on sources, levels and human exposure of hexachlorobenzene (HCB) in China. Potential sources of unintended HCB emission were assessed by production information, emission factors and environmental policies. HCB was observed in various environmental
Lesa L Aylward et al.
Regulatory toxicology and pharmacology : RTP, 58(1), 25-32 (2010-06-16)
Recent efforts worldwide have resulted in a growing database of measured concentrations of chemicals in blood and urine samples taken from the general population. However, few tools exist to assist in the interpretation of the measured values in a health

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service