Skip to Content
Merck
All Photos(3)

Key Documents

T55301

Sigma-Aldrich

2,4,6-Trichlorophenol

98%

Synonym(s):

1,3,5-Trichloro-2-hydroxybenzene, 2,4,6-TCP, 2,4,6-Trichlorophenol, BTS 45186, Dowicide 2S, Omal, Phenachlor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2OH
CAS Number:
Molecular Weight:
197.45
Beilstein:
776729
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 76.5 °C)

Quality Level

Assay

98%

bp

246 °C (lit.)

mp

64-66 °C (lit.)

SMILES string

Oc1c(Cl)cc(Cl)cc1Cl

InChI

1S/C6H3Cl3O/c7-3-1-4(8)6(10)5(9)2-3/h1-2,10H

InChI key

LINPIYWFGCPVIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Meiqin Hu et al.
Environmental science & technology, 46(16), 9005-9011 (2012-07-26)
Molecular iodine has been studied, for the first time, as a sensitizer for the degradation of 2,4,6-trichlorophenol (TCP) in aqueous solution under visible light (λ ≥ 450 nm). TCP was degraded in the presence of commercial I(2), but the reaction
Nadavala Siva Kumar et al.
Colloids and surfaces. B, Biointerfaces, 94, 125-132 (2012-03-01)
Biosorption of 2,4,6-trichlorophenol (2,4,6-TCP) from aqueous solution by biomass prepared from Acacia leucocephala bark, an agricultural solid waste has been investigated in the present study. All the experiments are carried out by batch mode technique. The resulting biosorbent was characterized
Yabo Wang et al.
ACS applied materials & interfaces, 4(8), 3965-3972 (2012-07-12)
Cu(2)O/TiO(2) heterojunction photocatalyst is built on carbon aerogel (CA) substrate with good adsorption properties by sol impregnation and seed-mediated synthesis approach. The Cu(2)O/TiO(2)/CA electrode has excellent electrosorptive and high efficient photocatalytic properties. Its morphology and surface chemical composition are characterized
Minhee Kim et al.
Chemosphere, 89(3), 262-268 (2012-05-17)
The well-known cosolvency-induced sorption model is not applicable to predict the sorption of carboxylic acids in cosolvent system. To investigate the phenomenon, sorption and solubility of chlorinated phenols (2,4-dichlorophenol (2,4-DCP) and 2,4,6-trichlorophenol (2,4,6-TCP)) and carboxylic acids (benzoic acid and 2,4-dichlorophenoxyacetic
Aki Sinkkonen et al.
Environmental science and pollution research international, 20(3), 1737-1745 (2012-07-04)
Chlorophenols, like many other synthetic compounds, are persistent problem in industrial areas. These compounds are easily degraded in certain natural environments where the top soil is organic. Some studies suggest that mineral soil contaminated with organic compounds is rapidly remediated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service