Skip to Content
Merck
All Photos(1)

Key Documents

40008

Supelco

Hexachlorobenzene solution

certified reference material, 1000 μg/mL in acetone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

Quality Level

Agency

MISA (Canada) Group 22

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Clc1c(c(c(c(c1Cl)Cl)Cl)Cl)Cl

InChI

1S/C6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9

InChI key

CKAPSXZOOQJIBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lifei Zhanga et al.
Environmental technology, 33(16-18), 1945-1951 (2012-12-18)
Decomposition of hexachlorobenzene (HCB) on high surface area y-alumina was comprehensively studied using in situ Fourier transform infrared spectroscopy, gas chromatography/mass spectrometry (GC-MS) and ion chromatography. Monitoring of the temperature-dependent decomposition by Fourier transform infrared spectroscopy showed that HCB decomposed
Yuanqing Zhou et al.
Water research, 47(1), 102-110 (2012-10-24)
We studied the dechlorination of hexachlorobenzene (HCB) in wetland mesocosm (MC) trials filled with sediment (well mineralized homogenized peat mixed with mud) from a wastewater treatment wetland located in a floodplain: three MCs were planted with common reed (Phragmites australis)
Xiaoqin Nie et al.
Chemosphere, 90(9), 2403-2407 (2013-01-01)
Synthesized lead-iron (Pb/Fe) bimetallic particles were applied to dechlorinate hexachlorobenzene (HCB) under various conditions (e.g. bimetal amount, initial pH value, reaction temperature, and reaction duration). The results showed that adding Pb onto Fe benefited the dechlorination of HCB and the
Renée Dallaire et al.
Environment international, 54, 85-91 (2013-02-21)
Several studies have reported negative associations of polychlorinated biphenyls (PCBs), hexachlorobenzene (HCB) and mercury (Hg) with duration of gestation and fetal growth in fish eating populations. Docosahexaenoic acid (DHA) from fish, seafood and marine mammal intake has been reported to
Carolina Andrea Pontillo et al.
Toxicology and applied pharmacology, 268(3), 331-342 (2013-03-07)
Hexachlorobenzene (HCB) is a widespread organochlorine pesticide, considered a possible human carcinogen. It is a dioxin-like compound and a weak ligand of the aryl hydrocarbon receptor (AhR). We have found that HCB activates c-Src/HER1/STAT5b and HER1/ERK1/2 signaling pathways and cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service