Skip to Content
Merck
All Photos(1)

Key Documents

45418

Supelco

Daminozide

PESTANAL®, analytical standard

Synonym(s):

Succinic acid mono(2,2-dimethylhydrazide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH2CONHN(CH3)2
CAS Number:
Molecular Weight:
160.17
Beilstein:
1863230
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

162-164 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CN(C)NC(=O)CCC(O)=O

InChI

1S/C6H12N2O3/c1-8(2)7-5(9)3-4-6(10)11/h3-4H2,1-2H3,(H,7,9)(H,10,11)

InChI key

NOQGZXFMHARMLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Daminozide is a commonly used plant growth regulator, which acts by inhibiting 2-oxoglutarate-dependent dioxygenase activity in the gibberellin biosynthesis pathway.

Application

Daminozide may be used as a reference standard for the determination of daminozide in fruits and vegetables by liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS).
It may also be used as a reference standard for the determination of daminozide in Ophiopogon japonicus plants and soil by ultra-performance liquid chromatography-triple quadrupole tandem mass spectrometry (UPLC-QqQ-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid determination of polar pesticides and plant growth regulators in fruits and vegetables by liquid chromatography/tandem mass spectrometry.
Chamkasem N.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 8(2), 1-10 (2018)
Daminozide reduces red color intensity in herbaceous peony (Paeonia lactiflora Pall.) flowers by inhibiting the expression of flavonoid biosynthetic genes.
Tang Y, et al.
3 Biotech, 8(2), 102-102 (2018)
Synthesis of Gibberellic Acid Derivatives and Their Effects on Plant Growth.
Tian H, et al.
Molecules (Basel), 22(5), 694-694 (2017)
A simple multi-residue method for determination of plant growth retardants in Ophiopogon japonicus and soil using ultra-performance liquid chromatography-tandem mass spectrometry.
Zhao X, et al.
Chemosphere, 207(2), 329-336 (2018)
K T Faughnan et al.
Journal - Association of Official Analytical Chemists, 74(4), 682-692 (1991-07-01)
A modified version of the Conditt and Baumgardner gas chromatographic/mass spectroscopic (GC/MS) method for determination of daminozide in peanut butter and raw peanuts is described. Daminozide in the food product is hydrolyzed to unsymmetrical dimethylhydrazine (UDMH) by sodium hydroxide digestion.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service