Skip to Content
Merck
All Photos(1)

Key Documents

W504009

Sigma-Aldrich

Amyl acetate

≥99%, FG

Synonym(s):

Pentyl acetate, Amyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COO(CH2)4CH3
CAS Number:
Molecular Weight:
130.18
Beilstein:
1744753
EC Number:
Council of Europe no.:
211
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.021
NACRES:
NA.21
Organoleptic:
banana; ethereal; fruity
grade:
FG
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal

reg. compliance

EU Regulation 1334/2008 & 178/2002

vapor density

4.5 (vs air)

vapor pressure

4 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

680 °F
714 °F

refractive index

n20/D 1.402 (lit.)

bp

142-149 °C (lit.)

mp

−100 °C (lit.)

density

0.876 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

banana; ethereal; fruity

SMILES string

CCCCCOC(C)=O

InChI

1S/C7H14O2/c1-3-4-5-6-9-7(2)8/h3-6H2,1-2H3

InChI key

PGMYKACGEOXYJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amyl acetate is an aliphatic ester that occurs naturally in banana fruit. It may be one of the odor volatiles produced by fungi such as Aspergillus, Penicillium and Fusarium species.

Application

Amyl acetate is a fruity odor compound that can be used as an attractant (component) in nanoemulsion formulation used to trap fruit flies (Drosophila melanogaster).
Banana essence. Used as test odorant in studies of olfactory function and in studies of the psychosocial effects of odor.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

105.8 °F

Flash Point(C)

41 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Correlation of gas chromatographic data with flavor profiles of fresh banana fruit.
McCarthy AI, et al.
Journal of Food Science, 28(4), 379-384 (1963)
Michael Schleyer et al.
Learning & memory (Cold Spring Harbor, N.Y.), 18(10), 639-653 (2011-09-29)
Drosophila larvae combine a numerically simple brain, a correspondingly moderate behavioral complexity, and the availability of a rich toolbox for transgenic manipulation. This makes them attractive as a study case when trying to achieve a circuit-level understanding of behavior organization.
Oshrat Tzhayik et al.
Ultrasonics sonochemistry, 19(4), 858-863 (2012-01-26)
Protein microspheres have been prepared by sonicating a mixture of pure fragrant oil (amyl acetate (AA)) with an aqueous protein (bovine serum albumin) solution. The prepared protein spheres are nano- to micrometer sized with an encapsulation efficiency of approx. 97%
Vidyulata Kamath et al.
Schizophrenia bulletin, 39(1), 59-67 (2011-05-28)
There is increasing evidence that schizophrenia patients have difficulties in the hedonic appraisal of odors. In a prior study, we assessed olfactory hedonic perception birhinally and found that males with schizophrenia failed to attach the appropriate hedonic valence to a
Janice K Kiecolt-Glaser et al.
Psychoneuroendocrinology, 33(3), 328-339 (2008-01-08)
Despite aromatherapy's popularity, efficacy data are scant, and potential mechanisms are controversial. This randomized controlled trial examined the psychological, autonomic, endocrine, and immune consequences of one purported relaxant odor (lavender), one stimulant odor (lemon), and a no-odor control (water), before

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service