Skip to Content
Merck
All Photos(1)

Key Documents

46259

Supelco

1,1,2,2-Tetrachloroethane

analytical standard

Synonym(s):

Acetylene tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CHCl2CHCl2
CAS Number:
Molecular Weight:
167.85
Beilstein:
969206
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

5.8 (vs air)

vapor pressure

8 mmHg ( 20 °C)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.494 (lit.)

bp

147 °C (lit.)

mp

−43 °C (lit.)

density

1.586 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

ClC(Cl)C(Cl)Cl

InChI

1S/C2H2Cl4/c3-1(4)2(5)6/h1-2H

InChI key

QPFMBZIOSGYJDE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,1,2,2-Tetrachloroethane may be used as an extraction solution for the simultaneous determination of chloramphenicol and thiamphenicol in honey samples using dispersive liquid-liquid microextraction followed by high-performance liquid chromatography.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dispersive liquid--liquid microextraction followed by high-performance liquid chromatography as an efficient and sensitive technique for simultaneous determination of chloramphenicol and thiamphenicol in honey
Chen H, et al.
Analytica Chimica Acta, 632(1), 80-85 (2009)
Wan-Chun Tsai et al.
Journal of chromatography. A, 1216(45), 7846-7850 (2009-10-07)
A novel sample preparation method "Dispersive liquid-liquid-liquid microextraction" (DLLLME) was developed in this study. DLLLME was combined with liquid chromatography system to determine chlorophenoxy acid herbicide in aqueous samples. DLLLME is a rapid and environmentally friendly sample pretreatment method. In
Marcos Caroli Rezende et al.
Magnetic resonance in chemistry : MRC, 47(6), 505-510 (2009-04-01)
Spectra of the N-phenyl-5,6-dihydro-2,4-diphenylbenzo[h]quinolinium tetrafluoroborate (1) and of the N-phenyl-5,6,8,9-tetrahydro-7-phenyldibenzo[c,h]acridinium tetrafluoroborate (2) were recorded in various solvents and temperatures. The analysis of the (1)H-NMR spectra of the tetrafluoroborate salt 1, recorded in acetone, acetonitrile, 1,1,2,2-tetrachloroethane and chloroform, revealed the existence
Srigokul Upadhyayula et al.
The journal of physical chemistry. B, 115(30), 9473-9490 (2011-06-21)
Electrostatic properties of proteins are crucial for their functionality. Carboxyamides are small polar groups that, as peptide bonds, are principal structural components of proteins that govern their electrostatic properties. We investigated the medium dependence of the molar polarization and of
Martin Elsner et al.
Environmental science & technology, 41(11), 4111-4117 (2007-07-07)
Despite widespread implementation of zero-valent iron remediation schemes, the manner and order of chemical bond cleavage in iron-mediated organohalide transformations remains imperfectly understood. We present insights from carbon isotope fractionation for the dehalogenation of 1,1,2,2-tetrachloroethane (1,1,2,2-TeCA) and 1,1,1-trichloroethane (1,1,1-TCA) by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service