Skip to Content
Merck
All Photos(2)

Key Documents

35030

Supelco

Chlorthion

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9ClNO5PS
CAS Number:
Molecular Weight:
297.65
Beilstein:
2292513
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=S)(OC)Oc1ccc(c(Cl)c1)[N+]([O-])=O

InChI

1S/C8H9ClNO5PS/c1-13-16(17,14-2)15-6-3-4-8(10(11)12)7(9)5-6/h3-5H,1-2H3

InChI key

NZNRRXXETLSZRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Chlorthion is an organophosphorus pesticide which belongs to the class of phosphorothionates. It inhibits acetylcholinesterase enzyme activity.

Application

Chlorthion may be used as an analytical reference standard for the determination of the analyte in aqueous samples and fruits/vegetables by solid-phase microextraction(SPME) in-line coupled to gas chromatography-mass spectrometry(GC-MS), and modified quick, easy, cheap, effective, rugged and safe(QUECHERS) method with magnetic nanoparticles and gas chromatography tandem mass spectrometry, respectively.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of multiple pesticides in fruits and vegetables using a modified quick, easy, cheap, effective, rugged and safe method with magnetic nanoparticles and gas chromatography tandem mass spectrometry.
Li YF, et al.
Journal of Chromatography A, 1361(11), 77-87 (2014)
Determination of pesticides in aqueous samples by solid-phase microextraction in-line coupled to gas chromatography?mass spectrometry.
Eisert R and Levsen K
Journal of the American Society For Mass Spectrometry, 6(11), 1119-1130 (1995)
Analysis of the time-dependent acute aquatic toxicity of organophosphorus pesticides: The critical target occupation model.
Legierse, et al.
Environmental Science & Technology, 33(6), 917-925 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service