Skip to Content
Merck
All Photos(1)

Key Documents

45485

Supelco

Fenchlorphos

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8Cl3O3PS
CAS Number:
Molecular Weight:
321.55
Beilstein:
1885571
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=S)(OC)Oc1cc(Cl)c(Cl)cc1Cl

InChI

1S/C8H8Cl3O3PS/c1-12-15(16,13-2)14-8-4-6(10)5(9)3-7(8)11/h3-4H,1-2H3

InChI key

JHJOOSLFWRRSGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fenchlorphos is an organophosphorus insecticide, which is commonly used on sheep and cattle against a number of internal and external parasites.
Fenchlorphos is an organophosphorus insecticide, which is commonly used on sheep and cattle against a number of internal and external parasites.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of Fenchlorphos residues in milk.
Webley JD, et al.
Analyst, 86(1024) (1961)
Ovarian and testicular function in the blue fox (Alopex lagopus) after oral administration of fenchlorphos during the breeding season.
G N Berge et al.
Acta veterinaria Scandinavica, 24(2), 200-210 (1983-01-01)
[Histochemical studies on the effect of fenchlorphos (0,0-dimethyl-0-2,4,5-trichlophenyl thiophosphate) on perfused liver of albino rat].
B Sawicki et al.
Patologia polska, 32(1), 79-88 (1981-01-01)
K S Khera et al.
Journal of toxicology and environmental health, 10(1), 111-119 (1982-07-01)
Insecticidal formulations in the form of dusts or aerosols containing rotenone from the root of Derris elliptica and pyrethrum from the flower of Chrysanthemum cinerariaefolium are commonly used in homes and gardens. Ronnel (fenchlorphos), a systemic insecticide, is used to
T S Rumsey et al.
Journal of animal science, 53(1), 217-225 (1981-07-01)
Four feeding trials were conducted to determine (1) how the organophosphate ronnel affects thyroid and adrenal circulating hormone levels and blood profile measurements of beef steers and (2) whether the effects of ronnel observed in beef steers could be produced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service