Skip to Content
Merck
All Photos(1)

Documents

34038

Supelco

Isoeugenol

analytical standard

Synonym(s):

2-Methoxy-4-propenylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(CH=CHCH3)OH
CAS Number:
Molecular Weight:
164.20
Beilstein:
1909602
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

>1 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.575 (lit.)

bp

132 °C/10 mmHg (lit.)

density

1.084 g/cm3

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

OC1=CC=C(/C=C/C)C=C1OC

InChI

1S/C10H12O2/c1-3-4-8-5-6-9(11)10(7-8)12-2/h3-7,11H,1-2H3/b4-3+

InChI key

BJIOGJUNALELMI-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isoeugenol belongs to the class of phenylpropenes, naturally occurring in the essential oils of plants. It can also be obtained from eugenol by isomerization. It has been used as a substrate in the preparation of vanillin via enzymatic transformation by lipoxygenase enzyme.

Application

Isoeugenol may be used as an analytical reference standard for the determination of the analyte in bioconversion broth using reversed-phase high-performance liquid chromatography with ultraviolet detection (RP-HPLC-UV).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The monolignol pathway contributes to the biosynthesis of volatile phenylpropenes in flowers.
Muhlemann KJ, et al.
The New phytologist, 204(3), 661-670 (2014)
Determination of vanillin, eugenol and isoeugenol by RP-HPLC.
Li-H-Y, et al.
Chromatographia, 60(11-12), 709-713 (2004)
Sathya N Prasad et al.
Neurotoxicology, 33(5), 1254-1264 (2012-07-31)
Acrylamide (ACR) intoxication in its monomeric form leads to neuronal damage in both experimental animals and humans. Oxidative stress is one of the principle mechanisms related to the neurotoxicity of ACR exposure. Hence, the present study aimed to recapitulate the
Morahem Ashengroph et al.
Applied biochemistry and biotechnology, 166(1), 1-12 (2011-10-13)
To screen strains of halotolerant or halophile bacteria which are able to convert isoeugenol to vanillin, 36 different strains of bacteria isolated from the salty environments in Iran were investigated. During growth on isoeugenol, a moderately halotolerant Gram-negative coccobacil showed
J Krupa et al.
The journal of physical chemistry. B, 116(36), 11148-11158 (2012-07-26)
In situ photochemical transformations of monomers of 2-methoxy-4-(prop-1-enyl)phenol (isoeugenol) and 2-methoxy-4-(prop-2-enyl)phenol (eugenol) isolated in low temperature matrices were induced by tunable UV laser light, and the progress of the reactions was followed by FTIR spectroscopy. Conformer-selective E ↔ Z geometrical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service