Merck
  • Ultraviolet-tunable laser induced phototransformations of matrix isolated isoeugenol and eugenol.
Ultraviolet-tunable laser induced phototransformations of matrix isolated isoeugenol and eugenol.
The journal of physical chemistry. B (2012-07-26)
J Krupa, A Olbert-Majkut, I Reva, R Fausto, M Wierzejewska
ABSTRACT
In situ photochemical transformations of monomers of 2-methoxy-4-(prop-1-enyl)phenol (isoeugenol) and 2-methoxy-4-(prop-2-enyl)phenol (eugenol) isolated in low temperature matrices were induced by tunable UV laser light, and the progress of the reactions was followed by FTIR spectroscopy. Conformer-selective E ↔ Z geometrical isomerizations could be successfully induced by irradiation at different wavelengths from the 310-298 nm range in the isoeugenol molecule, contains an asymmetrically substituted exocyclic C═C bond. Photolysis of both studied compounds was also observed, with H-atom shift from the OH group and formation of two types of long-chain conjugated ketenes. The photoproduced ketenes were found to undergo subsequent photodecarbonylation. Interpretation of the observed photoprocesses was supported by quantum chemical calculations undertaken at different levels of theory (DFT, MP2, QCISD).
Product Description
Supelco
Isoeugenol, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Isoeugenol, mixture of cis and trans, 99%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Isoeugenol, 98%, mixture of cis and trans
Pricing and availability is not currently available.