Merck
Conversion of isoeugenol to vanillin by Psychrobacter sp. strain CSW4.
Applied biochemistry and biotechnology (2011-10-13)
Morahem Ashengroph, Iraj Nahvi, Hamid Zarkesh-Esfahani, Fariborz Momenbeik
ABSTRACT
To screen strains of halotolerant or halophile bacteria which are able to convert isoeugenol to vanillin, 36 different strains of bacteria isolated from the salty environments in Iran were investigated. During growth on isoeugenol, a moderately halotolerant Gram-negative coccobacil showed capability of converting isoeugenol to vanillin. Based on morphological, physiological, and phylogenetic studies, strain CSW4 was classified as a bacterium belonging to the genus Psychrobacter. The bioconversion products were confirmed by thin-layer chromatography, high-performance liquid chromatography, and spectral data obtained from UV/Vis spectroscopy, FTIR, and mass-spectroscopy. Using growing cells, vanillin reached its maximum level of 88.18 mg L(-1) after 24 h of reaction time in the presence of 1 g L(-1) isoeugenol, resulting in a molar yield of 10.2%. The use of resting cells led to the optimal yield of vanillin (16.4%) which was obtained after 18-h reaction using 1 g L(-1) isoeugenol and 3.1 g of dry weight of cells per liter harvested at the end of the exponential growth phase. To improve vanillin yield, the effect of substrate concentration on vanillin production under resting cells conditions was also investigated. Using 10 g L(-1) isoeugenol, the maximal vanillin concentration (1.28 g L(-1)) was achieved after a 48-h reaction, without further optimization. The present study brings the first evidence for biotransformation of isoeugenol to vanillin in the genus Psychrobacter.
Product Description
Supelco
Isoeugenol, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Isoeugenol, 98%, mixture of cis and trans
Pricing and availability is not currently available.
Sigma-Aldrich
Isoeugenol, mixture of cis and trans, 99%, FG
Pricing and availability is not currently available.