Skip to Content
Merck
All Photos(3)

Key Documents

330574

Sigma-Aldrich

Indophenol Blue

Dye content 60 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N2O
CAS Number:
Molecular Weight:
276.33
Colour Index Number:
49700
Beilstein:
2379761
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 60%

technique(s)

titration: suitable

mp

168-170 °C (lit.)

λmax

606 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CN(C)c1ccc(cc1)\N=C2/C=CC(=O)c3ccccc23

InChI

1S/C18H16N2O/c1-20(2)14-9-7-13(8-10-14)19-17-11-12-18(21)16-6-4-3-5-15(16)17/h3-12H,1-2H3/b19-17+

InChI key

VRZJGENLTNRAIG-HTXNQAPBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indophenol Blue is used to colorimetrically measure N-containing compounds. The free amino acids react to form a blue color complex.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Zhen Wu et al.
Environmental science and pollution research international, 25(31), 31307-31317 (2018-09-09)
Knowledge about the impacts of fresh and field-aged biochar amendments on greenhouse gas (CH4, N2O) emissions is limited. A field experiment was initiated in 2012 to study the effects of fresh and field-aged biochar additions on CH4 and N2O emissions
Jin Zhou et al.
Microbial ecology, 76(3), 592-609 (2018-02-15)
A central goal in marine microecology is to understand the ecological factors shaping spatiotemporal microbial patterns and the underlying processes. We hypothesized that abiotic and/or biotic interactions are probably more important for explaining the distribution patterns of marine bacterioplankton than
Optimizing Nitrogen Management in Food and Energy Production and Environmental Protection, 13-13 (2001)
Leticia M Mesa et al.
Archives of environmental contamination and toxicology, 75(2), 316-326 (2018-05-31)
Two anthelmintic macrocyclic lactones-ivermectin and moxidectin-have revolutionized parasite control in cattle. These drugs are only partly metabolized by livestock, and the main route of excretion is via feces. In seasonally inundated floodplains, cattle feces come into direct contact with surface
Luis Abdala-Roberts et al.
American journal of botany, 103(12), 2070-2078 (2016-12-15)
It is generally thought that herbivore pressure is higher at lower elevations where climate is warmer and less seasonal, and that this has led to higher levels of plant defense investment at low elevations. However, the generality of this expectation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service